Elution Buffer for Dynabeads™ mRNA Purification Kits
Invitrogen™

Elution Buffer for Dynabeads™ mRNA Purification Kits

This Elution Buffer (10 mM Tris-HCl, pH 7.5) is included in the following Dynabeads™ mRNA purification kits: Cat. Nos. 61006,Read more
Catalog NumberQuantity
A335661 x 15 mL
Catalog number A33566
Price (JPY)
-
Quantity:
1 x 15 mL
This Elution Buffer (10 mM Tris-HCl, pH 7.5) is included in the following Dynabeads™ mRNA purification kits: Cat. Nos. 61006, 61011, 61012, and 61021. It is made available separately for applications that require more Elution Buffer than is provided in the kit.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineDynabeads
Quantity1 x 15 mL
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
1 x 15 mL, store at 2°C to 8°C