MagMAX™ Cell-Free DNA Lysis/Binding Buffer
Applied Biosystems™
MagMAX™ Cell-Free DNA Lysis/Binding Buffer
This buffer is included in the MagMAX Cell-Free DNA Isolation Kit (Cat. No. A29319). It is made available separately forRead more
Have Questions?
Catalog NumberQuantity
A33600425 mL
Catalog number A33600
Price (USD)
1,384.00
Each
Quantity:
425 mL
Price (USD)
1,384.00
Each
This buffer is included in the MagMAX Cell-Free DNA Isolation Kit (Cat. No. A29319). It is made available separately for applications that require more Lysis/Binding Buffer than is provided in the kit.
NOTE: The color of this Lysis/ Binding solution can range from colorless to yellow. The color of the solution has no impact on DNA extraction performance.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Next-Generation Sequencing, Real-Time PCR
High-throughput CompatibilityHigh-throughput Compatible, Manual Protocols, Automated Protocols
Product LineMagMAX
Product TypeDNA Lysis/Binding Buffer
Quantity425 mL
Sample TypeBlood
Shipping ConditionRoom Temperature
TargetGenomic DNA
Unit SizeEach