Fetal Bovine Serum, dialyzed, US origin, One Shot™ format, 50 mL - FAQs
Fetal Bovine Serum, dialyzed, US origin, One Shot™ format, 50 mL - FAQs
View additional product information for Fetal Bovine Serum, dialyzed, US origin - FAQs (26400044, A3382001)
11 product FAQs found
What are the benefits and disadvantages of using heat-inactivated FBS in cell culture?
  1. Heating inactivates complement. Active complement can participate in cytolytic events, contract smooth muscle, release histamine from mast cells and platelets, and activate lymphocytic and macrophage cells. Applications where heat-inactivated serum is recommended include immunological studies and culturing of embryonic stem cells (ESCs), insect cells, and smooth muscle cells.
  2. Heat inactivation helps to achieve bottle-to-bottle and lot-to-lot stability by neutralizing many factors that can vary largely from lot to lot.
  3. There aren't necessarily disadvantages to heat inactivation of FBS, but there is some evidence that suggests there may be no added benefit to it unless you are carrying out immune studies.
Note: Heat inactivation is performed in a 56 degrees C water bath for 30 min with swirling every 10 min or so for heat distribution and to lower the degree of protein aggregation/flocculant precipitation. Note: If the time or temperature is exceeded, the serum may thicken to a gel. If this occurs, the serum is no longer usable. Unnecessary heat inactivation can take up time and potentially lead to wasted reagents if a mistake is made during the protocol1.
1. Pellerin, et al., Bioengineering, published in 2021.
Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.
Are proteins removed from Fetal Bovine Serum, dialyzed (Cat. No. 26400044, A3382001)?
The raw serum is processed through a dialysis system that has a 10 kDa cutoff. Therefore, many proteins, peptides, as well as some hormones, glucose, amino acids, and other exogenous materials are removed.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Are there any other packaging configurations available for the new One Shot FBS product?
Yes, we also offer a 10 X 50 mL package, which is equivalent to a 500 mL bottle of FBS.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How is the new One Shot FBS bottle different from our existing One Shot bottle?
Our previous One Shot bottle, a 60 mL Nalgene media bottle, is not designed for easy pipetting and pouring whereas the new One Shot FBS bottle has a propietary design that was developed specifically for FBS use. This design allows you to freeze, thaw and pour without the need to aliquot, which minimizes the amount of FBS handling and reduces the risk of contamination.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Does the new One Shot FBS bottle fit into a conical tube rack?
The new One Shot FBS bottle is not shaped to fit into a traditional conical tube rack. It does however fit nicely in a 30 mm test tube rack for the purposes of storage and use in a water bath and biological safety cabinet.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the best way to hold the new One Shot FBS bottle for pouring?
For optimal, burp-free pouring, securely grip the bottle using the indents and pour with the chamfered edge facing down.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the shelf life of the new One Shot FBS?
The initial shelf life/stability for the new One Shot FBS is two years from the date of manufacture. Additional updates will continue to be made as data becomes available.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can I heat inactivate the serum in the new One Shot FBS bottle?
Yes, the serum in the new One Shot FBS bottle can be heat inactivated at 56 degrees C for 30 minutes.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can I reuse the new One Shot FBS bottles?
It is at the discretion of the customer to reuse these bottles for purposes other than what they are intended for. But please note that they cannot be autoclaved.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What type of plastic is the new One Shot FBS bottle made from?
It is made from polyethylene terephthalate (PET, PETE). This is the same plastic used in our other bottles.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.