DMEM for SILAC
Thermo Scientific™

DMEM for SILAC

Analyze protein expression by mass spectrometry using these media or additives for stable isotope labeling with amino acids in cell culture (SILAC).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A338226 x 500 mL
88364500 mL
Catalog number A33822
Price (USD)
309.65
Online exclusive
356.00
Save 46.35
Each
Quantity:
6 x 500 mL
Price (USD)
309.65
Online exclusive
356.00
Save 46.35
Each
Thermo Scientific™ DMEM for SILAC is optimized for use with stable isotope labeling with amino acids in cell culture (SILAC) to analyze protein expression by mass spectrometry (MS).

Features of DMEM for SILAC:

  • Flexible—liquid media deficient in both L-lysine and L-arginine, allowing for more complete proteome coverage through multiple isotopic amino acid labeling
  • High quality—medium is sterile, endotoxin-free, and cell culture-compatible

SILAC is a powerful method to identify and quantify relative differential changes in complex protein samples. This approach entails the in vivo metabolic incorporation of 'heavy' 13C- or 15N-labeled amino acids into proteins followed by mass spectrometry (MS) for the accelerated and comprehensive identification, characterization, and quantitation of proteins.

This SILAC medium is used together with heavy and light amino acids for the differential isotopic labeling of cells for SILAC analysis. SILAC media supplemented with heavy amino acids are capable of nearly 100% label incorporation into proteins of living cells.

Related products

  • NeuCode™ Lysine -080, 50 mg (A33613)
  • Fetal Bovine Serum, dialyzed, US origin (26400036)
  • RPMI 1640 Media for SILAC (88365)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeLiquid Medium
For Use With (Equipment)Mass Spectrometer
Label or DyeMass Isotope of Amino Acid
Quantity6 x 500 mL
Shipping ConditionRoom Temperature
FormLiquid
Product TypeMedia
Starting MaterialCell Lysate
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.

Frequently asked questions (FAQs)

Does DMEM for SILAC (Cat. No. 88364, A33822) have the labeled amino acids in it or do those have to be added separately?

DMEM for SILAC (Cat. No. 88364, A33822) is a liquid medium deficient in both L-lysine and L-arginine. It is optimized for use with stable isotope labeling with amino acids in cell culture (SILAC) to analyze protein expression by mass spectrometry (MS).

Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.