Automated Thermal Cycler (ATC), 384-well
Applied Biosystems™

Automated Thermal Cycler (ATC), 384-well

For customers seeking to automate their PCR workflow, the Applied Biosystems Automated Thermal Cycler (ATC) provides the reliability and performance you’ve come to expect from Applied Biosystems instruments.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoDescripción
A33981Includes 3-meter connection cable
A33980Includes 1-meter connection cable
A33982Includes 10-cm connection cable
Número de catálogo A33981
Precio (MXN)
-
Descripción:
Includes 3-meter connection cable
For customers seeking to automate their PCR workflow, the Applied Biosystems Automated Thermal Cycler (ATC) provides the reliability and performance you’ve come to expect from Applied Biosystems instruments. Now in a small, easy-to-integrate, and compatible format, our Automated Thermal Cycler enables hands-free PCR results.

Space-saving design with our smallest thermal cycler footprint
• Flexible modular design helps save space on robotic deck
• Separate control panel module provides flexibility when integrating on robotic deck
• Choice of 10-cm, 1-m, and 3-m cables allows flexibility to locate control module under thermal cycler, under deck, or up to 3 meters away

Automated lid and cycler features
• Automated lid compatible with programming for easy, hands-free operation with a liquid handler or plate stacker
• Heated cover slides forward to cover plate nest, helping ensure thermal contact and eliminate evaporation
• Compatible with full-skirt or half-skirt plates

Fully integrate your automated system with our easy-to-install software
• Integrate with robot via SiLA Rapid Integration standard–compatible software and prewritten application programming interfaces (APIs) for major robotic platforms
• Test instrument using desktop software before buying robot
• Enable ATC to cycle like old instrument with Applied Biosystems 9700 system simulation modes

Use the ATC as a stand-alone PCR thermal cycler
Flexible software can be connected to your robot's software interface or used to control the ATC as a stand-alone instrument. Test your instrument using the ATC desktop application before integrating it into your robotic automation system.

Award-winning service and support
• Help reduce downtime with rapid exchange support
• Have our engineers perform temperature qualification at your site

Applied Biosystems PCR plastics
Applied Biosystems PCR plastics have been designed and validated to work with our thermal cyclers for more than 25 years. Options for every format and throughput need are available to ensure consistent, precise results for any experiment.

Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.

Especificaciones
DescripciónIncludes 3-meter connection cable
Dimensiones12.5 x 7.1 x 5.3 in. (31.7 x 17.9 x 13.3 cm)
Tipo de pantallaSin visualización
Compatibilidad de alto rendimientoDiseñado para flujos de trabajo robóticos de alto rendimiento
Velocidad de rampa máxima2,8 °C/s (bloque), 1,6 °C/s (muestra)
Cantidad1 system
Velocidad de reacciónEstándar
Exactitud térmica±0,25 °C (de 35 °C a 99,9 °C)
Intervalo térmicode 0°C a 105°C
Uniformidad térmica< 0,5 °C (20 segundos después de alcanzar 95 °C)
Interfaz de usuarioUSB, Wi-Fi, Ethernet
Tensión100/240 V
GarantíaGarantía limitada
Vataje600 w
Peso6 kg (13,2 lb)
Configuraciones de bloques384-well Isothermal Block
Capacidad1 x 384 pocillos con placa de faldón completo
Tipo de productoTermociclador
Unit SizeEach
Contenido y almacenamiento
con cable de 3m

Preguntas frecuentes

Do any of your thermal cyclers, other than the Automated Thermal Cycler, hold a full skirted PCR plate?

For full skirt plate, a raised heating block is required. None of our thermal cyclers have a raised heating block except the Automated Thermal Cycler.

Find additional tips, troubleshooting help, and resources within our PCR Thermal Cyclers Support Center.

What is the optimal ramp rate for my thermal cycler?

There is no universal ramp rate for a thermal cycler because both the block and sample ramp rates are different for each thermal cycler. The sample ramp rate changes with the volume of the sample since more time and energy are required to heat higher sample volumes. The ramp rate may also need to be changed for a particular PCR assay or primer pair if you are trying to use the same PCR assay on two different thermal cyclers, for example.

As a reference, the block ramp rate commonly used with DreamTaq Polymerase on the MiniAmp thermal cycler is 3.0 C/sec. In a 50 µL PCR reaction, this translates to a sample ramp rate of ~1 C/sec.

Please refer to the following application note for further information about thermocycler ramp rates: https://assets.thermofisher.com/TFS-Assets/LSG/brochures/Thermal_Cycler_Ramp_RatesAppNote.pdf

Find additional tips, troubleshooting help, and resources within ourPCR Thermal Cyclers Support Center.

How can I monitor my thermal cycler remotely without being on the Cloud?

The Thermal Cycler Fleet Control Software allows monitoring of the following instruments from within a firewall:
SimpliAmp Thermal Cycler
ProFlex PCR System
Veriti Thermal Cycler
VeritiPro Thermal Cycler
Automated Thermal Cycler (ATC)

Which thermocyclers can be controlled using the Thermal Cycler Fleet Control Software?

The Thermal Cycler Fleet Control Software is compatible with the following instruments:
- SimpliAmp Thermal Cycler
- ProFlex PCR System
- Veriti Thermal Cycler
- VeritiPro Thermal Cycler
- Automated Thermal Cycler (ATC)
- MiniAmp Thermal Cycler
- MiniAmp Plus Thermal Cycler