본 제품은 LMO 제품으로, 고객 분께서 LMO 신고 시스템을 통해 직접 수입 신고를 진행해주셔야 합니다. 자세히보기
CTS™ CytoTune™-iPS 2.1 Sendai Reprogramming Kit
Invitrogen™

CTS™ CytoTune™-iPS 2.1 Sendai Reprogramming Kit

The CTS CytoTune-iPS 2.1 Sendai Reprogramming Kit is the first off-the-shelf reprogramming system designed for clinical and translational research. It delivers efficient, integration-free reprogramming of somatic cells into induced pluripotent stem cells.
Have Questions?
카탈로그 번호수량
A345461 Kit
카탈로그 번호 A34546
제품 가격(KRW)
21,227,000
온라인 행사
Ends: 31-Dec-2026
23,585,000
할인액 2,358,000 (10%)
Each
수량:
1 Kit
제품 가격(KRW)
21,227,000
온라인 행사
Ends: 31-Dec-2026
23,585,000
할인액 2,358,000 (10%)
Each

The CTS CytoTune-iPS 2.1 Sendai Reprogramming Kit is the first off-the-shelf reprogramming system designed for clinical and translational research. It delivers efficient, integration-free reprogramming of somatic cells into induced pluripotent stem cells. Like the CytoTune-iPS 2.0 Sendai kit, the CTS CytoTune-iPS 2.1 Sendai kit uses Sendai particles to deliver Yamanaka factors that are critical for efficient generation of induced pluripotent stem cells (iPSCs). This kit includes three vector preparations: polycistronic Klf4–Oct3/4–Sox2 (KOS), L-Myc, and Klf4 and is sufficient to reprogram a minimum of six wells of a six-well plate.

Supports a xeno-free workflow for the generation of iPSC lines

The bovine serum albumin carrier protein was removed from the CTS CytoTune-iPS 2.1 Sendai vector formulation. The preparation contains no animal-derived components at the primary component level, enabling the xeno-free generation of iPSCs from both fibroblasts and blood.

Facilitates customer regulatory filings with extensive testing and traceability documentation

The CTS CytoTune-iPS 2.1 Sendai kit is manufactured at a site with a process that was audited for conformity with current Good Manufacturing Practices for medical devices, 21 CFR Part 820 of the regulation. Full traceability documentation such as Certificate of Origin and Certificate of Analysis are available.

사양
배달 유형Sendai
용도(애플리케이션)Cell and Gene Therapy Research, Development, and Manufacturing
제품라인CTS
제품 유형iPSC Reprogramming Kit
수량1 Kit
유통 기한24 months from date of manufacture
배송 조건Dry Ice
Unit SizeEach
구성 및 보관
• 0.2 mL CTS CytoTune KOS
• 0.2 mL CTS CytoTune Klf4
• 0.2 mL CTS CytoTune L-myc

Store at -70°C.

자주 묻는 질문(FAQ)

What is CTS?

The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.

How many transductions can I perform with one CTS CytoTune-iPS 2.1 Sendai Reprogramming Kit?

The CTS CytoTune-iPS 2.1 Sendai Reprogramming Kit is sufficient to reprogram a minimum of six wells in a six-well plate. One well has a surface area of approximately 10 cm2.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Are the reprogramming steps in the CTS CytoTune-iPS 2.1 Sendai Reprogramming Kit different compared to CytoTune-iPS 2.0 Sendai Reprogramming Kit?

No, the reprogramming steps are in principle the same but it is important to consider the difference in titer between the two kits when transducing cells at certain MOIs.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Why is the CTS CytoTune-iPS 2.1 Sendai Reprogramming Kit so much more expensive than the CytoTune-iPS 2.0 Sendai Reprogramming Kit?

It is more expensive because of its label use statement: For research use and non-commercial manufacturing of cell based products for clinical research. CAUTION: Not intended for direct administration into animals or humans. The kit is manufactured according to GMP requirements and the drug master file is available for submission to FDA upon customer request, for use in clinical research.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Why does the CTS CytoTune-iPS 2.1 Sendai Reprogramming Kit contain L-Myc virus instead of c-Myc virus?

The L-Myc virus was chosen over the c-Myc virus because it is considered superior from a safety perspective, for clinical applications (L-myc is reported to have lower transformation/oncogenic potential than c-myc).

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.