TaqMan™ Advanced miRNA Human Endogenous Controls Card
Applied Biosystems™

TaqMan™ Advanced miRNA Human Endogenous Controls Card

The Applied Biosystems TaqMan Advanced miRNA Human Endogenous Controls Card provides the convenience of pre-spotted TaqMan Advanced miRNA assays inRead more
Have Questions?
Catalog NumberQuantity
A34716384 reactions
Catalog number A34716
Price (USD)
556.00
Each
Quantity:
384 reactions
Price (USD)
556.00
Each

The Applied Biosystems TaqMan Advanced miRNA Human Endogenous Controls Card provides the convenience of pre-spotted TaqMan Advanced miRNA assays in a 384-well microfluidic card format. This product contains one card than can be used to evaluate 30 potential endogenous control miRNAs in triplicate for up to four samples. Two exogenous (non-human) controls are included for normalization of sample input.

Features of the TaqMan Advanced miRNA Human Endogenous Controls Card include:
• Reproducibility—pre-spotted assays help reduce pipetting error and minimize experimental variability
• Sample savings—requires only 2 μL purified plasma/serum or 1–10 ng total RNA per sample
• Updated content—assay content updated for mirBase v.22
• Easy to use—just add a mix of diluted miR-Amp and master mix and run the qPCR; no additional pipetting required
• Gold-standard TaqMan qPCR chemistry—TaqMan Advanced miRNA assays draw on Thermo Fisher Scientific's industry-leading bioinformatics assay design pipeline and verification system to help ensure high specificity and minimal cross-reactivity, even for closely-related miRNA families

Product details:

Number of cards: 1
Number of unique miRNAs (not including exogenous controls): 30
Exogenous controls: cel-miR-39-3p, ath-miR159a
Compatible with the following Applied Biosystems qPCR instruments: Viia7, QuantStudio 7 Flex, and QuantStudio 12k Flex
Recommended master mix: TaqMan Fast Advanced Master Mix

Notes
• TaqMan Advanced miRNA cards require the use of the TaqMan Advanced cDNA Synthesis Kit, which uses a universal reverse transcription (RT) chemistry to prepare the cDNA template for use with TaqMan Advanced miRNA assays for detection and quantification of mature human miRNAs in biological samples.
• These plates are not compatible with the first-generation TaqMan MicroRNA cDNA Synthesis Kit, which uses a miRNA-specific reverse transcription chemistry.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodPrimer-Probe Detection
For Use With (Application)miRNA Analysis
Label or Dye2 Color
No. of Reactions384 reactions
Product LineTaqMan
Product TypeEndogenous Controls Card
Quantity384 reactions
Reaction SpeedFast or Standard
Shipping ConditionApproved for shipment at Room Temperature or on Wet or Dry Ice
Control TypeEndogenous Control
Format384-well Microfluidic Card
RNAi TypemiRNA
SpeciesHuman
Unit SizeEach
Contents & Storage
One 384-well array

The TaqMan miRNA card is shipped at ambient temperature. Upon arrival, store at 2°C to 8°C and keep protected from light.

Frequently asked questions (FAQs)

I am not seeing good amplification with my TaqMan Advanced miRNA array cards. What is wrong?

The TaqMan Advanced miRNA array cards allow one to profile over 300 different miRNA targets on one card. Due to the large number of targets and the nature of your sample, it is possible that some miRNA on the card will not be expressed on your sample, or expressed at very low levels. This is one reason why you will not see good amplification plots for every spot on the array card. Another possibility is the sample quality or input. Make sure to check the concentration and quality of the RNA sample before reverse transcription. Check the Ct values for your endogenous control assays on the card, and see if it is in the expected range for your typical samples.