TMT11-131C Label Reagent, 1 x 5 mg - FAQs
TMT11-131C Label Reagent, 1 x 5 mg - FAQs
View additional product information for TMT11-131C Label Reagent - FAQs (A37725, A37724, A34808, A34807)
1 product FAQs found
Can the TMT and TMTpro reagents be mixed?
No, the TMT and TMTpro reagents cannot be mixed because the reporter ions are in the same region.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.