What is the advantage of using Calf Serum with Iron (US Origin)?
The addition of iron in calf serum can result in higher performance of serum in cell culture applications. Iron-supplemented calf serum can be a viable alternative to fetal bovine serum in the growth of certain cells. Calf Serum with Iron (US Origin) is collected from calves 1-6 months of age, which naturally have higher levels of transferrin to bind iron. The iron content in Calf Serum with Iron (US Origin) is 500-700 µg/dL.