Invitrogen™

Attune™ NxT Automation SmartStor Sample Management System

Catalog number: A35221
Invitrogen™

Attune™ NxT Automation SmartStor Sample Management System

Catalog number: A35221
Catalog Number
A35221
Unit Size
1 system
Price (USD)
Catalog NumberUnit SizePrice (USD)
A352211 system
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recommendations
The Attune NxT Automation SmartStor Sample Management System is an accessory optimized for use in an automation line with the Attune NxT Acoustic Focusing Cytometer, autosampler, and Orbitor microplate mover. The SmartStor system’s presentation nest on the left-hand side allows the Orbitor arm to pick and load plates onto the autosampler. The SmartStor system maintains temperature from 4 to 40°C and holds up to 20 standard-height microplates. A 9-position hotel for deep-well plates is available for separate purchase (Cat. No. A38244). The SmartStor system comes with a 20-plate stack that is easily removed and replaced with the 9-position hotel if storage of deep well plates is needed.

Specifications

Block Format
Non-Interchangeable
Shipping Condition
Room Temperature
For Use With (Equipment)
Attune™ NxT Acoustic Focusing Cytometer
Product Line
Attune™
Product Type
Flow Cytometer Sample Management System

Figures