Thermo Scientific™

iBright™ FL1000 Imaging System Extended Warranty Package

Catalog number:  A35286

 Related applications: Sanger Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Invitrogen iBright FL1000 Imaging System is a powerful, easy-to-use western blot and gel documentation system. It provides sensitive, streamlined, multimode image capture and connects to the user through a large capacitive touch-screen interface and intelligently designed software, providing stunningly easy western blot and gel imaging. Combining an extended warranty with your system purchase, help extend the life of your investment. This package includes the system, SmartStart, and an extended warranty.

Key features of the iBright FL1000 Imaging System include:
• Flexible imaging options—capture visible and near-infrared fluorescence-based western blots and gels, chemiluminescent western blots, and colorimetric stained protein and DNA gels
• Smart Exposure-enabled advanced acquisition—designed to deliver high quality images every time with auto-alignment, auto exposure, auto-zoom, and auto-focus
• Advanced, intuitive interface—un-box, set up, and image in minutes with streamlined workflows
• On-board analysis with automated image analysis with multi-frame capability—analyze multiple mini gels or blots at once
• Small footprint—all capabilities are packed into a conveniently sized bench top instrument with a large imaging area for imaging multiple blots or gels at one time
• Thermo Fisher Cloud-enabled—a secure place to store, analyze, and share data

For detailed instrument information and specifications, see the iBright FL1000 Imaging System.

Get up and running fast with SmartStart
SmartStart is performed by a Field Application Scientist in your lab. SmartStart helps you:
• Get up and running quickly
• Get connected to your local network and the Cloud
• Choose appropriate reagents and consumables to meet your needs

Content will vary depending on your level of knowledge and experience.

Extended Warranty Plan
Add additional coverage to your existing manufacturer’s limited warranty with an extended warranty. Each extended warranty includes a planned maintenance visit. Should your instrument require a repair, the extended warranty coverage includes parts, labor, and travel at no additional cost. In addition, you’ll receive:
• On-site guaranteed 2-day response time*
• Scheduled on-site planned maintenance (PM)
• Priority access to Remote Service Engineers

Certain exclusions apply. Contact your Services and Support Representative for details at service.sales@thermofisher.com.

*Availability limited in some geographical areas.

Package components

Unit size

Qty

iBright™ FL1000 Imaging System Catalog number:A32752 1 system 1
A34016 Catalog number:A34016 1
ZG61SCIBRIGHTFL Catalog number:ZG61SCIBRIGHTFL 3
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: Chemiluminescent, IR Fluorescence, Nucleic acid gels, RGB Fluorescence
Dimensions: 68.0 x 38.2 x 59.8 cm
Display Interface: 12.1-inch, Capacitive touch screen
Field: 22.5 x 18.0 cm
Includes: Green LED, Visible LED, near-IR LED
Number of Channels: 7 positions motorized
Plan Duration: 3 Year Extended Warranty
Product Line: iBright™
Product Size: System + 3 yr ext warranty
Service Plan Response Time: 48 hours (availability limited in some geographic areas)
Services Included: Engineer travel and labor, Planned maintenance visit, Priority access to applications and instrument technical support teams, Priority access to off-site remote service engineer, Replacement service spare parts
Services Not Included: Consumables, Instrument accessories, Instrument and software upgrades, Reagents
Shipping Condition: Room Temperature
Weight: 50 kg (110 lbs)

Documents

Manuals & protocols