TrueGuide™ sgRNA Negative Control, non-targeting 1
Invitrogen™
TrueGuide™ sgRNA Negative Control, non-targeting 1
TrueGuide Synthetic sgRNA Negative Control does not recognize any sequence in the human or mouse genome and provides a non-edited experimental control.
Have Questions?
카탈로그 번호수량
A355263 nmol
카탈로그 번호 A35526
제품 가격(KRW)
351,000
온라인 행사
Ends: 30-Sep-2026
390,000
할인액 39,000 (10%)
Each
수량:
3 nmol
제품 가격(KRW)
351,000
온라인 행사
Ends: 30-Sep-2026
390,000
할인액 39,000 (10%)
Each
TrueGuide negative controls are non-targeting gRNA sequences that do not recognize any sequence in the human or mouse genome and provide a non-edited control when performing analysis on pooled populations of edited cells.
Non-targeting gRNA sequences that do not recognize any sequence in the human or mouse genome and provide a non-edited control when performing analysis on pooled populations of edited cells.
Validated negative control crRNA. Universal control for Mouse and Human
For Research Use Only. Not for use in diagnostic procedures.
사양
제품라인TrueGuide
제품 유형gRNA
수량3 nmol
배송 조건Room Temperature
제어 유형Negative Control
형식Tube
RNAi TypegRNA
Human, Mouse
Unit SizeEach
구성 및 보관
Delivered as dried RNA. Store dried RNA at room temperature; store resuspended RNA at -5°C to -30°C. Resuspend in nuclease-free water or TE.
자주 묻는 질문(FAQ)
I would like to calculate the molecular weight of TrueGuide Synthetic gRNA? What is the length of TrueGuide Synthetic gRNA (TrueGuide sgRNA)?
The length of TrueGuide sgRNA is typically 100 nt. Also, the exact molecular weight can also be found on the COA.