Product Image
Applied Biosystems™
96-well Septa for SeqStudio™ Genetic Analyzer
Se trata de septos de placa de 96 pocillos para su uso en el analizador genético SeqStudio.
Have Questions?
Número de catálogoCantidad
A3564120 septos
Número de catálogo A35641
Precio (MXN)
-
Cantidad:
20 septos
These are 96-well plate septa for use on the SeqStudio Genetic Analyzer.
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Cantidad20 septos
Condiciones de envíoTemperatura ambiente
Línea de productosSeqStudio
TipoSeptos del analizador genético
Unit SizeEach
Preguntas frecuentes
What does the SeqStudio Starter Kit (PN A35000) contain?
The SeqStudio Starter Kit (PN A35000) includes:
- SeqStudio Integrated Capillary Protector (Cat. No. A31923)
- SeqStudio Cathode Buffer Container (Cat. No. A33401)
- Sequencing Standard, BigDye Terminator v3.1, for 3500/SeqStudio Flex (Cat. No. 4404312)
- DS-33 GeneScan Installation Standards with GeneScan 600 LIZ Size Standard v2.0 (Cat. No. 4376911)
- Hi-Di Formamide (Cat. No. 4401457)
- MicroAmp Optical 96-Well Reaction Plate with Barcode (Cat. No. 4306737)
- MicroAmp 96-Well Tray/Retainer Set (Cat. No. 403081)
- MicroAmp Optical 8-Tube Strip, 0.2 mL (Cat. No. 4316567)
- Nuclease-Free Water (not DEPC-Treated) (Cat. No. AM9930)
- Cathode Buffer Container Reservoir Septa for SeqStudio Genetic Analyzer (Cat. No. A35640)
- 96-well Septa for SeqStudio Genetic Analyzer (Cat. No. A35641)
- 8-strip Septa for SeqStudio Genetic Analyzer (Cat. No. A35643)
- SeqStudio Cartridge v2 (PN A41328) (if this component is ordered online the catalog number is A41331)
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.
How many times can I reuse the 96-well Septa for SeqStudio Genetic Analyzer?
The 96-well Septa for SeqStudio Genetic Analyzer are intended for single use only. We recommend replacing the plate septa after each run. If rinsed and reused, the septa can become sticky and gummy over time and may start sticking to the array. This may potentially increase the risk of damaging the array.
Bleach, detergents, or solvents should not be used to clean the septa as this can cause them to dry out faster and risk contaminating the system.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.
How often can the plate septa on the 3130/xl, 3730/xl, 3500/xl, and SeqStudio instruments be used?
We officially recommend that the plate septa be used one time only. However, we are aware that some of our customers will rinse the septa with water and re-use them. In those instances, the septa can become sticky and gummy over time and eventually start sticking to the array. In the best case scenario, the array will lift the plate off of the autosampler just enough to break the connection with the plate sensor and abort the run. The Event Log might show that the run aborted due to the user aborting it, so the run will appear to have completed or stopped with no errors and no data. The worst case scenario is that the plate remains on the array as the autosampler starts to move, bending the electrodes and breaking the array. We recommend replacing the plate septa regularly (replacing after each run is recommended). Do not use bleach, detergents, or solvents to clean the septa, it will cause them to dry out faster and also risks contaminating the system.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.