Oncomine™ Breast cfDNA Research Assay v2 - FAQs
Oncomine™ Breast cfDNA Research Assay v2 - FAQs
View additional product information for Oncomine™ Breast cfDNA Research Assay v2 - FAQs (A35865)
17 product FAQs found
Can I use the Oncomine Breast cfDNA Assay v2 for BRCA testing?
Use the Oncomine Breast cfDNA Assay v2 (Cat. No. A35865) when your research requires sensitive detection of tumor-derived variants in cell-free DNA from plasma across multiple breast cancer-related genes (i.e., AKT1, CCND1, EGFR, ERBB2, ERBB3, ESR1, FBXW7, FGFR1, KRAS, PIK3CA, SF3B1, and TP53). This assay does not enable detection of the BRCA1/2 genes. For customers interested in evaluating the BRCA 1/2 genes from FFPE tissue or whole blood, the Oncomine BRCA Assay (Cat. No. A47912) enables accurate and sensitive detection of BRCA1/2 somatic and germline mutations.
How many Oncomine Breast cfDNA Research Assay v2 samples can I run on Ion 530, Ion 540, and Ion 550 chips?
For the Oncomine Breast cfDNA Research Assay v2, the Ion 530 Chip supports up to 5 libraries and the Ion 540 Chip supports up to 20 libraries. The Ion 550 Chip is not listed for use with this assay but customers may choose to independently test assays that are not supported.
Is the Oncomine Breast cfDNA Research Assay v2 compatible only with Ion Torrent systems?
Yes. The Oncomine Breast cfDNA Research Assay v2 is designed for the Ion Torrent workflow and is intended for use with Ion Torrent sequencing platforms rather than other NGS systems.
Where can I download the latest BED file for the Oncomine Breast cfDNA Research Assay v2?
The latest BED files can be downloaded from the Oncomine Breast cfDNA Research Assay v2 product page at https://www.thermofisher.com/order/catalog/product/A35865.
I created a planned Oncomine Breast cfDNA Research Assay v2 run in my Torrent Suite Software but cannot see it on my Ion GeneStudio S5 Series system. Why is this?
It is possible that there is a communication problem between the Ion GeneStudio S5 Series system and the server. We recommend shutting down the system, waiting for 2 minutes and then rebooting. If that does not work and if you are using the Ion S5 XL System, we recommend turning off the server and the system for 2 minutes. Then, attach a keyboard and monitor to the server and turn the server back on. Log in to the server and make sure you can connect to it via your web browser. Then, turn on the system. If the planned run does not appear, please contact Technical Support at techsupport@thermofisher.com for further assistance.
Why don't I see the Oncomine options in my Torrent Suite Software when planning an Oncomine Breast cfDNA Research Assay v2 run?
You would need to use Torrent Suite Software version 5.2 or later in order to see Oncomine options.
Can I run the Oncomine Breast cfDNA Research Assay v2 on the Ion S5 System or an Ion GeneStudio S5 Series system with a 510 or 520 chip?
The Oncomine Breast cfDNA Research Assay v2 has only been validated on the Ion 530 and 540 chips at this time.
Can I run the Oncomine Breast cfDNA Research Assay v2 on the Ion Proton System with a PI chip?
The Oncomine Breast cfDNA Research Assay v2 has only been validated on the Ion 530 and 540 chips at this time.
Can I run the Oncomine Breast cfDNA Research Assay v2 on the Ion PGM System using a 314, 316, or 318 chip?
The Oncomine Breast cfDNA Research Assay v2 has only been validated on the Ion 530 and 540 chips at this time.
Can I use a spectrophotometer to quantify my Oncomine Breast cfDNA Research Assay v2 library?
Library quantitation with a spectrophotometer may not be accurate as it will measure all DNA present instead of just the samples with both barcoded adapters attached.
The Oncomine Breast cfDNA Research Assay v2 was left at 4 degrees C for more than an hour after target amplification. Can I still use it?
The PCR products can be stored at 4 degrees C for up to one hour.
Can I use single-cell DNA in the Oncomine Breast cfDNA Research Assay v2?
No. The recommended input amount is 20 ng of cfDNA but successful libraries have been made from 1-50 ng of cfDNA.
Why can't I use a spectrophotometer to quantitate my cfDNA samples for the Oncomine Breast cfDNA Research Assay v2?
We do not recommend using spectrophotometer methods for quantifying cfDNA because they are not reliable with low concentrations of nucleic acids. Use of these methods can result in overestimation of the concentration of sample, under-seeding of the target amplification reaction, and low library yields.
What is the best way to quantitate my cfDNA samples for use in the Oncomine Breast cfDNA Research Assay v2?
We recommend using the Qubit dsDNA HS Assay Kit (Cat. No. Q32851) for quantifying the cfDNA samples.
Which DNA isolation kit do you recommend for the Oncomine Breast cfDNA Research Assay v2?
We recommend using the MagMAX Cell-Free DNA Isolation Kit (Cat. No. A29319), which was included in-house during the Oncomine cfDNA workflow verification testing. Please follow the “Alternate protocol for isolation of higher concentration cfDNA” found in Appendix B of the MagMAX Cell-Free DNA Isolation User Guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0014327_A29319_magmax_cellfree...lasms_ug.pdf).
Where can I find the sequences of the primers used in the Oncomine Breast cfDNA Research Assay v2?
The primer sequences for the Oncomine Breast cfDNA Research Assay v2 are proprietary.
What genes are covered in the Oncomine Breast cfDNA Research Assay v2?
The Oncomine Breast cfDNA Research Assay v2 covers the following genes:
- ALT1
- ERBB3
- KRAS
- CCND1
- ESR1
- PIK3CA
- EGFR
- FBXW7
- SF3B1
- ERBB2
- FGFR1
- TP53
More information on the coverage and the difference between versions 1 and 2 of the assay can be found in the flyer: Liquid Biopsy Cell Free Research Assays (https://assets.thermofisher.com/TFS-Assets/CSD/Flyers/liquid-biopsy-cell-free-research-assays-flyer.pdf).