Attune™ NxT Blocker Bar Conversion Kit - FAQs
Attune™ NxT Blocker Bar Conversion Kit - FAQs
View additional product information for Attune™ NxT Blocker Bar Conversion Kit - FAQs (A35966)
1 product FAQs found
Can I install the Attune Nxt Blocker Bar myself?
No. The installation of the Attune Nxt Blocker Bar must be done on site by a Themo Fisher Scientific engineer.