High Select™ Depletion Spin Columns
High Select™ Depletion Spin Columns
High Select™ Depletion Spin Columns
Thermo Scientific™

High Select™ Depletion Spin Columns

Decrease albumin, immunoglobulins, and other high-abundance proteins from human serum or plasma in preparation for mass spectrometry analysis, or 1D and 2D gel electrophoresis using High Select depletion spin columns or resin.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityProduct TypePurification Target
A363656 ColumnsMini Spin ColumnHSA/Immunoglobulin
A3636624 ColumnsMini Spin ColumnHSA/Immunoglobulin
A3636710 ColumnsMidi Spin ColumnHSA/Immunoglobulin
A3636850 mLResinHSA/Immunoglobulin
A3637110 ColumnsMidi Spin ColumnTop14 Abundant Protein
A3637250 mLResinTop14 Abundant Protein
A363696 ColumnsMini Spin ColumnTop14 Abundant Protein
A3637024 ColumnsMini Spin ColumnTop14 Abundant Protein
Catalog number A36365
Price (BRL)
2.165,06
Each
Quantity:
6 Columns
Product Type:
Mini Spin Column
Purification Target:
HSA/Immunoglobulin
Price (BRL)
2.165,06
Each
Analysis of human fluids is often complicated by the presence of high concentrations of serum proteins, including albumin and IgG that can make up more than 70% of total serum protein. Removal of albumin, immunoglobulins, and other abundant proteins is essential for the study of low-abundance proteins. Thermo Scientific High Select HSA/Immunoglobulin and Top 14 Abundant Protein depletion spin columns and resins enable depletion of the most abundant proteins in human serum and plasma for improved detection of low-abundance proteins using mass spectrometry analysis or 1D or 2D gel electrophoresis.
• Flexible—resin provided in mini and midi column formats as well as bulk for custom filling options
• Optimized—removes >95% of IgG and >95% of albumin (HSA/Immunoglobulin); removes >95% of IgG and >95% of albumin, plus 12 other abundant proteins (Top 14)
• Fast—process samples in ∼10 minutes
• Economical—cost-effective spin columns are priced for single use
• Reduced carry-over—one-time use prevents protein carryover and experimental variability

High Select HSA/Immunoglobulin Depletion Resin uses highly specific, immobilized anti-HSA and anti-immuoglobulin antibodies (IgG, IgA, IgM, IgD, and IgE) to remove human serum albumin (HSA) and all major subclasses of immunoglobulins from serum and plasma. High Select Top14 Abundant Protein Depletion Resin uses highly specific immobilized antibodies to remove human serum albumin (HSA), albumin, IgG, IgA, IgM, IgD, IgE, kappa and lambda light chains, alpha-1-acidglycoprotein, alpha-1-antitrypsin, alpha-2-macroglobulin, apolipoprotein A1, fibrinogen, haptoglobin, and transferrin from serum, plasma, or spinal fluids. The resins are provided in an economical and convenient spin column format specifically designed for one-step processing and single use or bulk format for custom filling.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeProtein
For Use With (Equipment)Mass Spectormeter
Label or DyeUnlabeled
Purification TargetHSA/Immunoglobulin
Quantity6 Columns
Shipping ConditionWet Ice
Workflow StepProtein Depletion
FormatMini Spin Column
Product LineHigh Select
Product TypeMini Spin Column
Starting MaterialPlasma, Serum
Unit SizeEach
Contents & Storage
Store at 2–8°C.

Frequently asked questions (FAQs)

Can I use the High Select HSA/Immunoglobulin Depletion Resin to process mouse serum samples?

No, the High Select HSA/Immunoglobulin Depletion Resin cannot be used for mouse samples; the depletion resin contains immobilized antibodies that only recognize human serum or plasma proteins.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Can I reuse the High Select HSA/Immunoglobulin Depletion Resin?

We don’t recommend reusing the High Select HSA/Immunoglobulin Depletion Resin as depletion efficiency may decrease after regeneration, and subsequent depleted samples may be contaminated from prior ones.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What formats of the High Select HSA/Immunoglobulin Depletion Resin do you offer?

The High Select HSA/Immunoglobulin Depletion Resin is offered in 3 formats:

a) Resin alone: High Select HSA/Immunoglobulin Depletion Resin (Cat. No. A36368)
b) High Select HSA/Immunoglobulin Depletion Mini Spin Columns (Cat. Nos. A36365, A36366) that can process a maximum of 10 µL of human serum or plasma
c) High Select HSA/Immunoglobulin Depletion Midi Spin Columns (Cat. No. A36367) that can process a maximum of 100 µL of human serum or plasma. For serum samples containing abnormally high amounts of albumin or IgG, the sample load may need to be reduced.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.