Product Image
Invitrogen™

Attune™ NxT Violet 6 Conversion Kit

The Attune NxT Violet 6 Conversion kits are designed to reconfigure Attune NxT four-channel violet instruments to allow detection of six channels of violet.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
A36569Violet
A35428Blue
A36571Red
A36572Yellow
Catalog number A36569
Price (USD)
-
Color:
Violet

Attune NxT Violet 6 Conversion kits are for use in the reconfiguration of four-channel Attune NxT instruments to enable detection of six channels of violet. A conversion kit is required for each original and any upgraded lasers of the instrument. The conversion is carried out in your laboratory by a Field Service Engineer.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorViolet
Product LineAttune
Quantity1 kit
Shipping ConditionRoom Temperature
For Use With (Equipment)Attune NxT Four-Channel Violet Instrument
Product TypeConversion Kit
Unit SizeEach
Contents & Storage
Optical block and filters