MagMAX™ DNA Multi-Sample Ultra 2.0 Kit - FAQs
MagMAX™ DNA Multi-Sample Ultra 2.0 Kit - FAQs
View additional product information for MagMAX™ DNA Multi-Sample Ultra 2.0 Kit - FAQs (A36570)
22 product FAQs found
I want to isolate genomic DNA from buccal cells. How do I do this?
We would recommend using one of our MagMAX DNA Multi-Sample Ultra 2.0 Kit or ChargeSwitch gDNA Buccal Cell Kits, which were designed to isolate high-quality genomic DNA from buccal samples. We also offer normalized kits, which will normalize isolated DNA concentration from sample to sample.
What will happen if the MagMAX HMW DNA Booster is not added during the workflow?
The isolated DNA recovered would likely not achieve greater than 100 kb in fragment size and the workflow reproducibility of the HMW kit would be compromised. If the customer wishes to leave the Booster Solution out of the workflow, we recommend following the guidance of Applied Biosystems(TM) MagMAX(TM) DNA Multi-Sample Ultra 2.0 kit to achieve genomic DNA without expanding into the HMW (>100 bp) or Ultra-HMW DNA (around 300 bp) recovery range.
If I have already purchased the MagMAX DNA Multi-Sample Ultra 2.0 Kit or standalone components, what do I need to extract high molecular weight DNA?
In addition to the MagMAX DNA Mult-Sample Ultra 2.0 product(s) you will need:
MagMAX HMW DNA Booster, 100 preps (Cat. No. A36624) or
In addition to the MagMAX DNA Mult-Sample Ultra 2.0 product(s) you will need:
MagMAX HMW DNA Booster, 100 preps (Cat. No. A36624) or MagMAX HMW DNA Booster, 1,000 preps (Cat No. A36625).
For cell and tissue samples include:
MagMAX Cell and Tissue DNA Extraction Buffer, 100 preps (Cat. No. A45469) or MagMAX Cell and Tissue DNA Extraction Buffer, 1,000 preps (Cat. No. A45470)
The MagMAX HMW DNA User Guide (MAN1001478)
The specific HMW DNA KingFisher automation protocol for your KingFisher Purification System (see product description page for download).
MagMAX HMW DNA Booster, 1,000 preps (Cat No. A36625).
For cell and tissue samples include:
MagMAX Cell and Tissue DNA Extraction Buffer, 100 preps (Cat. No. A45469) or MagMAX Cell and Tissue DNA Extraction Buffer, 1,000 preps (Cat. No. A45470)
The MagMAX HMW DNA User Guide (MAN1001478)
The specific HMW DNA KingFisher automation protocol for your KingFisher Purification System (see product description page for download).
Can the MagMAX HMW DNA Booster be paired with other genomic DNA isolation kits?
The MagMAX HMW DNA Booster is specifically designed and optimized for use with the MagMAX DNA Multi-Sample Ultra 2.0 (Cat. No. A36570) chemistry.
What is included in the MagMAX HMW DNA Kit?
The MagMAX HMW DNA Kit (Cat. No. A36626) includes: MagMAX DNA Multi-Sample Ultra 2.0 Kit (Cat. No. A36570), MagMAX HMW DNA Booster, 100 preps (Cat. No. A36624).
MagMAX HMW DNA Kit with Cell and Tissue DNA Extraction Buffer (Cat. No. A26627) includes: MagMAX DNA Multi-Sample Ultra 2.0 Kit (Cat. No. A36570), MagMAX Cell and Tissue DNA Extraction Buffer (Cat. No. A45469), MagMAX HMW DNA Booster, 100 preps (Cat. No. A36624).
What DNA extraction kit would you recommend for fragments smaller than 250 kb that are compatible with a KingFisher Flex Purification System?
We recommend the MagMAX DNA Multi-Sample Ultra 2.0 Kit (Cat. No. A36570) that is optimized for use on the moderate-throughput KingFisher Duo Prime Purification System (Cat. No. 5400110) and the higher-throughput KingFisher Flex Purification System, KingFisher with 96 Deep-well head (Cat. No. 5400630).
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.
What is the difference between Proteinase K for MagMAX DNA Multi-Sample Ultra Kit (Cat. No A25561) and Proteinase K for MagMAX 96 DNA Multi-Sample Kit (Cat. No. 4489110)?
The difference between Proteinase K for MagMAX‐96 DNA Multi‐Sample Kit and Proteinase K for MagMAX DNA Multi‐Sample Ultra Kit is the brand. The former is from Invitrogen legacy and the latter is from Applied Biosystems legacy. Both products have an identical concentration of 100 mg/mL and can be used interchangeably.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center
Are the DNA Binding Beads included within the MagMAX DNA Multi-Sample Ultra 2.0 Binding Beads (Cat. No. A36570) available as a stand-alone product and in a smaller volume?
We offer the MagMAX DNA Multi-Sample Ultra 2.0 Binding Beads (Cat. No. A36579), but the volume is 45 mL. The volume of the beads in the MagMAX DNA Multi-Sample Ultra 2.0 Kit (Cat. No. A36570) is 4.5 mL.
What is the level of detection for somatic variants with the Oncomine Myeloid Research Assay GX?
Detection of somatic variants with the Oncomine Myeloid Research Assay GX has been verified down to 5% allele frequency.
Which extraction kits are compatible with Oncomine Myeloid Research Assay GX?
Here is a list of kits that are compatible with Oncomine Myeloid Research Assay GX:
Nucleic acid isolation - RNA samples
- MagMAX mirVana Total RNA Isolation Kit, Cat. No. A27828
- PureLink Total RNA Blood Kit, Cat. No. K156001
- RNaseZap RNase Decontamination Solution, Cat. No. AM9780, AM9782, AM9784
Nucleic acid isolation - DNA samples
- MagMAX DNA Multi-Sample Ultra 2.0 Kit, Cat. No. A36570
- PureLink Genomic DNA Mini Kit, Cat. No. K1820-01
What are the sample input requirements for Oncomine Myeloid Research Assay GX?
The recommended minimum sample input is 27.75 ng of DNA (1.11 ng/µL minimum concentration) and 14.25 ng of RNA (0.95 ng/µL minimum concentration).
What sample controls should I use with the Oncomine Myeloid Research Assay GX?
The recommended controls are:
DNA Controls:
- Seraseq® Myeloid Mutation DNA Mix, Cat. No. 0710-0408 (SeraCare®)
- Myeloid DNA Reference Standard, Cat. No. HD829 (Horizon™)
- AcroMetrix™ Oncology Hotspot Control, Cat. No. 969056
RNA Controls:
- Universal Human Reference RNA, Cat. No. 740000 (Agilent™)
- Seraseq® Myeloid Fusion RNA Mix, Cat. No. 0710-0407 (SeraCare®)
How many Oncomine Myeloid Research Assay GX samples can be multiplexed on a GX5 Chip?
Multiplex sequencing of up to 8 Oncomine Myeloid Research Assay GX samples (8 DNA+ 8 RNA) per lane on a GX5 Chip can be performed in a single run. There are 4 lanes on the GX5 Chip, so 32 total samples (32 DNA+ 32 RNA) can be run on one GX5 Chip (8 samples per run).
What are the components of Oncomine Myeloid Research Assay GX?
The Oncomine Myeloid Research Assay GX (Cat. No. A47857) includes the 2-pool DNA panel, the 1-pool RNA panel, Genexus Strip 1, and Genexus Strip 2-AS. The DNA panel is provided in two packs of 8 tubes (4 tubes of Myeloid DNA Pool 1 and 4 tubes of Myeloid DNA Pool 2 per pack). The RNA panel is provided in one pack of 8 tubes (Myeloid RNA Pool 1). Each primer pool in the panel is provided in pairs of tubes, where each tube pair contains one tube with primers in position 1 and one empty uncapped tube in position 2. Three 8-strip packs of the Genexus Strip 1 and Genexus Strip 2-AS (24 total of each strip) are provided with each kit.
How many samples can I run using Oncomine Myeloid Research Assay GX?
The contents of each Oncomine Myeloid Research Assay GX kit are sufficient for up to 32 samples (32 x 3‑pool reactions: 32 x 2‑pool DNA reactions and 32 x 1‑pool RNA reactions).
How many primer pools are present in the Oncomine Myeloid Research Assay GX?
The Oncomine Myeloid Research Assay GX includes 3 pools of Ion AmpliSeq oligonucleotide primers (a 2-pool DNA panel and a 1-pool RNA panel).
What is the Oncomine Myeloid Research Assay GX gene content?
The Oncomine Myeloid Research Assay GX targets key genes and fusions associated with major myeloid disorders, including acute myeloid leukemia (AML), myeloid dysplastic syndrome (MDS), myeloproliferative neoplasms (MPN), chronic myeloid leukemia (CML), chronic myelomonocytic leukemia (CMML), and juvenile myelomonocytic leukemia (JMML). The DNA panel comprises 40 key genes while the RNA panel includes a broad fusion panel of 29 driver genes, enabling detection over 600 unique fusion isotypes. See flyer for gene targets (https://assets.thermofisher.com/TFS-Assets/CSD/Flyers/Oncomine_myeloid_assay_gx_flyer.pdf).
What is the Oncomine Myeloid Research Assay GX designed for ?
The Oncomine Myeloid Research Assay GX is a comprehensive targeted next-generation sequencing (NGS) assay designed for sensitive detection of myeloid disorder-associated DNA mutations and fusion transcripts in blood and bone marrow samples.
Can the MagMAX DNA Multi-Sample Ultra 2.0 Kit (Cat. No. A36570) be used to isolate genomic DNA from bone marrow aspirates and PBMCs?
Yes, we have protocols for using the MagMAX DNA Multi-Sample Ultra 2.0 Kit (Cat. No. A36570) to isolate genomic DNA from these sample types, but they require purchase of an additional Cell and Tissue Extraction Buffer. The kit and buffer can be purchased together as Cat. No. A45721 or the Cell and Tissue Extraction Buffer can be purchased as a stand alone item (Cat. No. A45469 or A45470).
The suggested workflow for bone marrow aspirates is as follows:
- Use only up to 200 µL of bone marrow aspirate
- Centrifuge at 200 x g for 10 mins
- Remove the supernatant (leaving about 50 µL of aspirate with the loose pellet) and resuspend the cell pellet in 400 µL of Cell and Tissue Extraction Buffer
- From this point, follow the protocol for isolation of DNA from buffy coat samples
The suggested workflow for 1-2.0 x 10E6 PBMCs is as follows:
- Centrifuge at 500-800 x g for 10 mins
- Remove the supernatant (leaving the cell pellet) an resuspend the cell pellet in 400 L of Cell and Tissue Extraction Buffer
- From this point, follow the protocol for isolation of DNA from cultured cell samples
Can the MagMAX DNA Multi-Sample Ultra 2.0 Kit (Cat. No. A36570) be used to isolate DNA from cultured cells and tissue?
Yes, we have protocols for using the MagMAX DNA Multi-Sample Ultra 2.0 Kit (Cat. No. A36570) to isolate DNA from cells and tissue, but they require purchase of an additional Cell and Tissue Extraction Buffer. The kit and buffer can be purchased together as Cat. No. A45721 or the Cell and Tissue Extraction Buffer can be purchased as a stand alone item (Cat. No. A45469 or A45470).
Do the MagMAX DNA Multi-Sample Ultra 2.0 cell and tissue DNA isolation protocols require an RNase A treatment step?
RNase A digestion is optional, but we highly recommend doing it. We recommend using PureLink RNase A (20 mg/mL) (Cat. No. 12091021).
Saliva samples can be processed with the MagMAX DNA Multi-Sample Ultra 2.0 Kit as well as MagMAX Saliva gDNA Isolation Kit. What is the difference between these kits and which one should I use?
Both kits use different magnetic beads and different buffers. Both kits can be used with stabilized and fresh saliva samples, but if you are exclusively working with saliva samples, we recommend using the MagMAX Saliva gDNA Isolation Kit. This kit has a faster and more optimized workflow for isolating genomic DNA from saliva samples.