MagMAX™ HMW DNA Kit with Cell and Tissue Extraction Buffer, up to 100 preps - FAQs
MagMAX™ HMW DNA Kit with Cell and Tissue Extraction Buffer, up to 100 preps - FAQs
View additional product information for MagMAX™ HMW DNA Kit and Booster - FAQs (A36627, A36626, A36625, A36624)
12 product FAQs found
What KingFisher Purification System accessories are needed to automate the HMW DNA isolation workflow?
For the KingFisher Flex automated workflows and KingFisher Apex automated workflows, customers will require the 96 deep-well magnetic head and corresponding 96 deep well heat block. For the KingFisher Duo Prime automated workflows, customers will require the 12-pin magnet head for 96 deep-well plate formats and the corresponding KingFisher Duo Prime heat block
KingFisher Sample Purification System Accessories.
What will happen if the MagMAX HMW DNA Booster is not added during the workflow?
The isolated DNA recovered would likely not achieve greater than 100 kb in fragment size and the workflow reproducibility of the HMW kit would be compromised. If the customer wishes to leave the Booster Solution out of the workflow, we recommend following the guidance of Applied Biosystems(TM) MagMAX(TM) DNA Multi-Sample Ultra 2.0 kit to achieve genomic DNA without expanding into the HMW (>100 bp) or Ultra-HMW DNA (around 300 bp) recovery range.
Why is it recommended to use a mixture of wide bore and standard P1000 pipette tips for the MagMAX HMW DNA workflow?
Fragile samples can get damaged when they are forced through the narrow orifice of standard pipette tips. Using a combination of standard and wide bore pipette tips helps to increase the yield of HMW DNA, keeping the fragment sizes intact. Reference the MagMAX HMW DNA User Guide (MAN1001478).
Are low-binding tubes essential for manual workflows?
Yes, low-bind tubes are recommended to minimize loss due to absorption to the tube walls. Please refer to MagMAX HMW DNA User Guide (MAN1001478) for guidance on when to transfer to new tubes to reduce risk to sample contamination.
Why does the MagMAX HMW DNA User Guide specify a step-by-step reagent addition when preparing the sample plate for blood and cell samples, but recommends a master mix (pre-prepared mixture of reagents) for tissues?
The enhancer solution improves proteinase K activity at optimal concentrations; however, excessive enhancer solution can reduce proteinase K effectiveness. For blood and cell samples, adding the blood sample and cell lysate in the correct order dilutes the enhancer to achieve optimal proteinase K activity during the digestion step. For tissue samples, the tissue must be digested with proteinase K before using the tissue lysate for extraction. Preparing a master mix with Cell/Tissue extraction buffer ensures the enhancer solution is diluted to the concentration needed for optimal proteinase K activity in these samples.
Is it necessary to use all wells within the KingFisher 96 deep-well plate?
No, it is not necessary to fill all wells of a KingFisher 96 deep-well plate. If you are using less than the full-plate format, simply don't add any reagents to the unused wells for the isolation workflow.
Are there larger volume standalone reagent components available to scale up the MagMAX HMW DNA workflow?
Yes, the following larger volume standalone components are available for purchase separately:
- MagMAX HMW DNA Booster, 1,000 preps (1.0 mL) (Cat. No. A36625)
- MagMAX DNA Multi-Sample Ultra 2.0 Proteinase K (45 mL) (Cat. No. A36578)
- MagMAX DNA Multi-Sample Ultra 2.0 Binding Beads (45 mL) (Cat. No. A36579)
- MagMAX DNA Multi-Sample Ultra 2.0 Wash I Solution (1.1 L) (Cat. No. A36580)
- MagMAX DNA Multi-Sample Ultra 2.0 Lysis Binding Solution (450 mL) (Cat No. A36581)
- MagMAX DNA Multi-Sample Ultra 2.0 Elution Solution (120 mL) (Cat No. A36582)
- MagMAX DNA Multi-Sample Ultra 2.0 Enhancer Solution (45 mL) (Cat No. A36583)
- MagMAX Cell and Tissue DNA Extraction Buffer (600 mL) (Cat No. A45470)
Is RNase A treatment necessary when isolating HMW DNA from cells or tissue samples?
The RNase A treatment is optional but highly recommended for optimal results. RNAse A treatment helps to remove RNA and single stranded DNA, increasing the purity of double stranded DNA. We recommend using the PureLink RNase A (20 mg/mL) (Cat. No. 12091021).
Can I perform a manual and automated nucleic acid isolation using the same MagMAX HMW DNA Kit?
Yes, the MagMAX HMW DNA Kit has been optimized for manual nucleic acid isolation using the DynaMag-2 Magnet (Cat. No. 12321D) in addition to automated isolation using the KingFisher Duo Prime, Flex, or Apex Purification Systems.
What sample types and corresponding input amounts were developed with the MagMAX HMW DNA Kit workflow?
Whole blood: 200 µL, Cells: 1 million (1 x 106), Tissue: low DNA yield tissue (i.e., brain) 8 mg/ high DNA yield tissue (i.e., spleen) ~2 mg.
What is the expected fragment size profile achieved with the MagMAX HMW DNA Kit?
With the MagMAX HMW DNA Kit, you can expect greater than 70% of your DNA fragments to be above 40 kb, achieving a primary fragment peak larger than 100 kb. If a fragment size below 40 kb is desired, we recommend using the MagMAX DNA Multi-Sample Ultra 2.0 Kit (Cat. No. A36570).
What is included in the MagMAX HMW DNA Kit?
The MagMAX HMW DNA Kit (Cat. No. A36626) includes: MagMAX DNA Multi-Sample Ultra 2.0 Kit (Cat. No. A36570), MagMAX HMW DNA Booster, 100 preps (Cat. No. A36624).
MagMAX HMW DNA Kit with Cell and Tissue DNA Extraction Buffer (Cat. No. A26627) includes: MagMAX DNA Multi-Sample Ultra 2.0 Kit (Cat. No. A36570), MagMAX Cell and Tissue DNA Extraction Buffer (Cat. No. A45469), MagMAX HMW DNA Booster, 100 preps (Cat. No. A36624).