MagMAX™ Cell-Free Total Nucleic Acid Isolation Kit
Applied Biosystems™
MagMAX™ Cell-Free Total Nucleic Acid Isolation Kit
The MagMAX Cell-Free Total Nucleic Acid Isolation Kit is designed for enrichment of all nucleic acid species present in cell-freeRead more
Have Questions?
Catalog NumberQuantity
A367161 kit
Catalog number A36716
Price (MXN)
18,672.12
Each
Quantity:
1 kit
Price (MXN)
18,672.12
Each
The MagMAX Cell-Free Total Nucleic Acid Isolation Kit is designed for enrichment of all nucleic acid species present in cell-free biological samples such as plasma. The kit utilizes MagMAX magnetic-bead technology, ensuring reproducible recovery of high-quality nucleic acid that is suitable for a broad range of applications, including real-time PCR, digital PCR, and next-generation sequencing.
The magnetic bead–based purification format allows you to easily process a wide range of sample input volumes (1 mL to 6 mL). When used with the KingFisher Duo Prime or KingFisher Flex Magnetic Particle Processor, 6 to 24 plasma samples of 2-mL input volume can be isolated at once. Alternatively, samples can be processed manually with the use of a magnetic stand.
Features of the MagMAX Cell-Free Total Nucleic Acid Isolation Kit include:
• Automation-ready, phenol-free extraction
• Fast procedure allows 6 to 24 samples to be processed in 90 minutes or less when used with the KingFisher Duo Prime or KingFisher Flex Magnetic Particle Processor with a 24 Deep Well Head
• Flexible protocol accommodates sample volume inputs ranging from 1 mL to 6 mL of plasma
• No need for carrier RNA
• Elution volumes ranging from 15 μL to 60 μL
• Compatible with the Oncomine Lung cfTNA Research Assay
Go beyond cell-free DNA and get the total picture
Isolating and analyzing circulating cell-free total nucleic acid (cfTNA) from plasma and other cell-free samples serves as a non-invasive method to detect and monitor targets associated with certain disease states such as cancer.
The first step in attaining this valuable information is efficient nucleic acid isolation that specifically recovers the circulating nucleic acid species that contains information related to the disease state. The importance of cell-free DNA (cfDNA) is becoming established, but it may not tell the whole story. Cell-free RNA (cfRNA) sequences serve as critical biomarkers as well in the form of microRNAs and gene fusions. This inclusion of the cfRNA with the cfDNA provides an opportunity for researchers to expand their studies and advance human health.
The MagMAX Cell-Free Total Nucleic Acid kit is developed by the same Thermo Fisher Scientific scientists that brought you the MagMAX Cell-Free DNA Isolation Kit. The difference is the inclusion of all cfRNA species in the eluate, allowing you to increase the amount of information available from a single cell-free sample. Just like the cfDNA Isolation Kit, the cfTNA kit is designed for both manual and automated isolation of nucleic acid from a wide sample volume input range of 1 to 6 mL. The phenol-free, magnetic-bead format makes the kit fully amenable to automated isolation.
Thanks to the scalable format that allows for various sample inputs, it is possible to vary the number of samples processed with a single kit. For example, enough reagents are provided to process 50 samples of 2 mL plasma each. Alternatively, with the same kit it is possible to process 25 samples of 4 mL plasma each. Higher and lower input volumes are possible and the number of samples that can be processed will vary accordingly. This unique product attribute allows you to tailor the product to your specific needs.
Benefits of MagMAX magnetic bead–based purification
Magnetic beads offer many benefits compared to other technologies for isolating nucleic acid. Beads bind the DNA more efficiently than glass-fiber filters, resulting in higher and more consistent yields. Additionally, because filters and vacuum manifolds are not used, there is no risk of filter clogging due to cellular particulates in samples. This clogging issue is of particular concern with protein-rich, large-volume samples such as plasma that are commonly used in circulating cell-free nucleic acid studies.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Next-Generation Sequencing, Real-Time PCR
High-throughput CompatibilityHigh-throughput Compatible, Manual Protocols, Automated Protocols
Product LineMagMAX
Product TypeTotal Nucleic Acid Isolation Kit
Quantity1 kit
Sample TypePlasma
Shipping ConditionRoom Temperature
Test Time90 min.
FormatKit
Isolation TechnologyMagnetic Bead
Unit SizeEach
Contents & Storage
• 175 mL MagMAX Cell-Free Total Nucleic Acid lysis/binding solution
• 100 mL (final volume) MagMAX Cell-Free Total Nucleic Acid wash solution concentrate
• 21 mL MagMAX Cell-Free Total Nucleic Acid elution solution
• 3.5 mL MagMAX Cell-Free Total Nucleic Acid magnetic beads
• 1.5 mL MagMAX Cell-Free Total Nucleic Acid proteinase K
Frequently asked questions (FAQs)
For the Oncomine Lung Cell-Free Total Nucleic Acid Research Assay, what are the analysis workflows in Ion Reporter Software?
There are two analysis workflows available:
1. Oncomine TagSeq Lung v2 Liquid Biopsy - w2.0 - Single Sample: Detects and annotates low-frequency variants including SNPs/Indels (down to 0.1% limit of detection), Fusions, and CNVs from targeted nucleic acid libraries (DNA & RNA) from the Oncomine Lung Cell‑Free Total Nucleic Acid Research Assay. This is compatible with DNA & RNA purified from cell-free total nucleic acids.
2. Oncomine TagSeq Lung v2 Tumor - w2.0 - Single Sample: Detects and annotates low-frequency variants including SNPs/Indels (down to 0.5% limit of detection), fusions, and CNVs from targeted nucleic acid libraries (DNA & RNA) from the Oncomine Lung Cell-Free Total Nucleic Acid Research Assay. Due to deamination events caused by the FFPE process, the minimum alternative allele frequency is set to 0.3%. This makes it compatible with DNA & RNA purified from FFPE tumor tissue as well as fresh frozen tumor tissue.
Are there any BED files for the Oncomine Lung Cell-Free Total Nucleic Acid Research Assay?
Yes, there is a specific BED file and Hotspot file for the Oncomine Lung Cell-Free Total Nucleic Acid Research Assay. Please contact your local Field Application Specialist (FAS) or Clinical Application Consultant (CAC) to request the BED files.
What is the LOD (limit of detection) for the Oncomine Lung Cell-Free Total Nucleic Acid (cfNA) Research Assay?
Through the use of Tag Sequencing technology, low limits of detection (LOD) can be achieved for different variant types*:
- For SNVs/short indels, an LOD of 0.1% can be achieved with sensitivity of ˜90% and specificity of >99%
- For fusions & MET exon skipping, an LOD of 1% can be achieved with sensitivity of >90% and specificity of >99%
- For MET CNV target, detection as low as 1.2-fold amplification can be achieved with sensitivity of >90% and specificity of >99%
*Sensitivity and specificity for each variant type were determined using a collection of contrived positive samples and cfNA isolated from normal healthy donors.
Does the Oncomine Lung Cell-Free Total Nucleic Acid Research Assay use AmpliSeq technology?
No, this assay uses Tag Sequencing technology. Cell-free DNA (cfDNA) and cell-free RNA (cfRNA) are found at extremely low concentrations in the plasma fraction of whole blood. Because of this low prevalence, Taq Sequencing technology is utilized in this assay. The technology attaches unique molecular tags to the gene-specific primers. After amplification, the tagged molecules are grouped based on the tags. Groups containing the same mutant variant 80% of the time or greater will be called positive. Using the Tag technology, groups that contain random errors generated through the library construction/sequencing process are removed.
Which kit should I use to isolate cell-free total nucleic acid (cfNA) for the Oncomine Lung Cell-Free Total Nucleic Acid Research Assay?
Use cell-free total nucleic acid (cfNA) extracted using a method optimized for cfNA isolation from plasma. We recommend the MagMAX Cell Free Total Nucleic Acid Isolation Kit (Cat. No. A36716). You can expect 5-50 ng of cfDNA and 5-100 pg of cfRNA from 10 mL blood research sample collected in a K2 EDTA blood collection tube (Cat. No. ??)