Pierce™ Anti-DYKDDDDK Affinity Resin
Thermo Scientific™

Pierce™ Anti-DYKDDDDK Affinity Resin

Pierce Anti-DYKDDDDK Spin Columns provide a fast, convenient method for purification of DYKDDDDK-tagged proteins expressed in in vitro protein expression systems, bacteria, yeast, and mammalian cells.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A36804100 mL
A3680310 mL
A368012 mL
A400045625 Columns
Catalog number A36804
Price (USD)
8,383.65
Online exclusive
9,420.00
Save 1,036.35
Each
Quantity:
100 mL
Price (USD)
8,383.65
Online exclusive
9,420.00
Save 1,036.35
Each

Thermo Scientific Pierce Anti-DYKDDDDK Spin Columns provide a fast, convenient method for purification of DYKDDDDK-tagged proteins expressed in in vitro protein expression systems, bacteria, yeast, and mammalian cells. The amino acid sequence DYKDDDDK, commonly known as 'FLAG', is recognized by a high-affinity rat monoclonal antibody (clone L5) that is covalently attached to UltraLink Biosupport, which is a rigid, hydrophilic, highly crosslinked, copolymeric, and porous resin with high coupling capacity.

For protein purification, resin is added to a sample containing DYKDDDDK-tagged proteins with the tag on either the N- or C-terminus. Following capture of DYKDDDDK-tagged proteins, non-specifically bound proteins can be washed away before dissociating bound target proteins with elution buffer.

Features of Pierce Anti-DYKDDDDK Affinity Resin

  • Specific—unique base beads and highly specific antibody minimize off-target binding (low non-specific binding)
  • High purity—optimized bind-wash-elute protocol enables high purity
  • High yield—special antibody conjugation method enables high yield
  • Rapid—entire purification protocol typically takes less than 40 mins
  • Economical—purification protocol allows multiple reuses
  • Flexible—available in multiple formats including bulk loose resin and spin columns

Characteristics of Pierce Anti-DYKDDDDK Affinity Resin

  • Composition: anti-DYKDDDDK antibody covalently attached to UltraLink Biosupport, a highly crosslinked rigid support
  • Particle size: 50–80 μm
  • pH stability: 1–13
  • Operating temperature: 2–30°C; do not freeze
  • Particle concentration: 50% slurry in phosphate buffered saline, 0.02% sodium azide, pH 7.5
  • Binding capacity: ≥3.0 mg DYKDDDDK-tGFP-His protein (∼32 kDa/mL settled resin)

Available formats

  • Resin slurries—2 mL, 10 mL, and 100 mL of 50% slurry in PBS with 0.02% sodium azide, pH 7.5
  • Spin columns—10 mL spin columns containing 3 mL of settled resin, supplied at 6 mL 50% slurry
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormLiquid
FormatLiquid
Formulation50% suspension in PBS containing 0.02% Sodium azide
Product LinePierce
Product TypeAffinity Resin
Quantity100 mL
Shipping ConditionWet Ice
Stationary PhaseAnti-DYKDDDDK monoclonal antibodies
MatrixAffinity Resin
Unit SizeEach
Contents & Storage
50 mL settled resin. 100 mL supplied as a 50% slurry in PBS containing 0.02% sodium azide. Store at 2–8°C.

Frequently asked questions (FAQs)

What is the slurry/suspension percentage for your anti-DYKDDDDK ("FLAG") products?

Anti-DYKDDDDK Magnetic Agarose (Cat. Nos. A36797, A36798, A36799B) is offered as a 25% suspension (1 mL of 25% suspension = 0.25 mL of settled beads). UltraLink-based Anti-DYKDDDDK Affinity Resin (Cat. Nos, A36801, A36803, A36804) is offered as a 50% slurry (1 mL of 50% slurry = 0.5 mL of settled resin).

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What is the binding capacity for your Anti-DYKDDDDH Affinity Resin?

Here is the binding capacity: ≥3.0 mg DYKDDDDK-tGFP-His protein (˜32 kDa)/mL settled resin.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Can I use your UltraLink-based Anti-DYKDDDDH ("FLAG") Affinity Resin for FPLC purification?

Yes. The UltraLink-based Anti-DYKDDDDH (“FLAG”) Affinity Resin (Cat. Nos. A36801, A36803, A36804) can be used for FPLC purification.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

How do I cleave off the DYKDDDDK ("FLAG") tag after purification?

An enterokinase cleavage site behind the DYKDDDDK (“FLAG”) tag can allow complete removal of the DYKDDDDK (“FLAG”) tag leaving no additional amino acids. We offer EKMax Enterokinase (Cat. Nos. E18001 and E18002) that can be used for this purpose. Subsequently, the EKMax Enterokinase can be removed using EK-Away Resin (Cat. No. R18001), a resin conjugated with soybean trypsin inhibitor, which has high affinity for enterokinase.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Citations & References (1)

Citations & References
Abstract
NLRP6 potentiates PI3K/AKT signalling by promoting autophagic degradation of p85α to drive tumorigenesis.
Authors:Zhi F,Li B,Zhang C,Xia F,Wang R,Xie W,Cai S,Zhang D,Kong R,Hu Y,Yang Y,Peng Y,Cui J
Journal:Nature communications
PubMed ID:37770465
The PI3K/AKT pathway plays an essential role in tumour development. NOD-like receptors (NLRs) regulate innate immunity and are implicated in cancer, but whether they are involved in PI3K/AKT pathway regulation is poorly understood. Here, we report that NLRP6 potentiates the PI3K/AKT pathway by binding and destabilizing p85α, the regulatory subunit ... More