CTS™ OpTmizer™ T-Cell Expansion SFM, no phenol red, bag format
Gibco CTS OpTmizer T-Cell Expansion SFM supports the growth and expansion of human T lymphocytes. It is a complete serum-free,Leia mais
Have Questions?
Número do catálogo
Quantity
A3705003
1000 mL
Número do catálogo A3705003
Preço (BRL)
-
Quantity:
1000 mL
Gibco CTS OpTmizer T-Cell Expansion SFM supports the growth and expansion of human T lymphocytes. It is a complete serum-free, xeno-free medium consisting of CTS OpTmizer T-Cell Expansion Basal Medium and CTS OpTmizer T-Cell Expansion Supplement, which are mixed together prior to use.
CTS OpTmizer T-Cell Expansion SFM is available with and without phenol red. This version of CTS OpTmizer SFM lacks phenol red, and is available in both bottle and bag format.
Features of CTS OpTmizer T-Cell Expansion SFM include: • Supports high-density T cell culture (>4 x 106 CD3+ T-cells/mL) • Supports T-cell activation using Dynabeads magnetic beads, soluble antibodies, and stimulatory antibody-presenting cell protocols • Similar phenotype, function (e.g., cytokine secretion profile), and viability to T-cells cultured with conventional human AB serum-supplemented medium • Supports a T-cell phenotype similar to human serum-supplemented medium • Demonstrates enhanced efficacy and persistence of CART-19 cells when grown in medium supplemented with CTS Immune Cell Serum Replacement (ICSR)
Demonstrated functional performance Cells grown in CTS OpTmizer T-Cell Expansion SFM with or without human serum demonstrate good cytokine production profiles in polyclonally activated T cells. CTS OpTmizer T-Cell Expansion SFM maintains the desired the CD62L+ central memory T-cell phenotype.
GMP manufactured The primary manufacturing site for CTS OpTmizer T-Cell Expansion SFM is FDA-registered and has an ISO13485-certified quality management system. The methods and controls used for the manufacturing are in conformity with current Good Manufacturing Practices for medical devices, 21 CFR Part 820, of the regulation. Alternate manufacturing sites are available, providing risk mitigation by helping to ensure security of supply.
Versatility CTS OpTmizer T-Cell Expansion SFM has been widely adopted in clinical T-cell research applications including the culture of CAR T-cells. It works with multiple culture formats, including culture flasks, culture bags, and rocking bioreactor systems. CTS OpTmizer T-Cell Expansion SFM was designed to support expansion of cultured human T-cells without additional serum supplementation. Increased performance is generally observed when supplemented with CTS Immune Cell Serum Replacement.
What you get CTS OpTmizer T-Cell Expansion SFM includes two components: 1 liter of CTS OpTmizer T-Cell Expansion Basal Medium and 26 mLs of CTS OpTmizer T-Cell Expansion Auxiliary Concentrated Medium, which are mixed together to create the complete medium. Full traceability documentation such as Certificate of Origin and Certificate of Analysis are available.
Especificações
Cell TypeT Cells
Product LineOpTmizer
Product TypeSerum Free Medium (SFM)
Quantity1000 mL
Shipping ConditionRoom Temperature
Without AdditivesNo Phenol Red
Unit SizeEach
Conteúdo e armazenamento
• 1 x 1000 mL bag CTS OpTmizer Expansion Basal Medium, store at 2°C to 8°C, protect from light • 1 x 26 mL CTS OpTmizer Expansion Supplement, store in the dark (at 2°C to 8°C.)
Frequently asked questions (FAQs)
What is CTS?
The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.
Is there a difference in formulation of the CTS OpTmizer T Cell Expansion SFM between the bottle and bag packaging formats?
The bottle and bag packaging formats of the medium have the same formulation.
Can I use CTS OpTmizer T Cell Expansion SFM for culture and expansion of mouse T cells? If not, which medium would you recommend?
We have not tested this medium on mouse T cells. If you're looking for a serum-free medium to culture mouse T cells, you can try AIM V Medium (Cat. No. 12055083 or 12055091). For mouse cells, we recommend adding 50 µM beta-mercaptoethanol (Cat. No. 21985023).
Can GlutaMAX be used instead of Glutamine to supplement CTS OpTmizer T-Cell Expansion SFM?
GlutaMAX cannot replace Glutamine because cells may not initially use GlutaMAX so that T cells cannot grow. We recommend adding 4 mM GlutaMAX and 2 mM Glutamine (as stated in the manual) into the CTS OpTmizer T-Cell Expansion SFM if GlutaMAX is needed.
What is the shelf life of CTS OpTmizer T-Cell Expansion SFM?
The complete CTS OpTmizer T-Cell Expansion SFM is stable for 4 weeks when stored in the dark at 2-8 degrees C. However, the two individual medium components are stable for 18 months when stored in the dark at 2-8 degrees C.