CTS™ OpTmizer™ T-Cell Expansion SFM, no phenol red
Gibco™
CTS™ OpTmizer™ T-Cell Expansion SFM, no phenol red
Gibco™ CTS OpTmizer T-Cell Expansion SFM supports the growth and expansion of human T lymphocytes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A37050055 L
A370500710 L
Catalog number A3705005
Price (EUR)
1 782,00
Each
Quantity:
5 L
Price (EUR)
1 782,00
Each
Gibco™ CTS OpTmizer T-Cell Expansion SFM supports the growth and expansion of human T lymphocytes. It is a complete serum-free, xeno-free medium consisting of CTS OpTmizer T-Cell Expansion Basal Medium and CTS OpTmizer T-Cell Expansion Supplement, which are mixed together prior to use.
CTS OpTmizer T-Cell Expansion SFM in 5 L and 10 L bag format is compatible with closed-system processing for media preparation. 5 L and 10 L volumes support scaling of cell therapy manufacturing workflows. The updated bag format allows for easier integration with standard cell therapy equipment, including CTS devices (Xenon, Rotea, and DynaCellect).
Features of CTS OpTmizer T-Cell Expansion SFM include:
  • Both kit components (medium and supplement) in Aegis5-14 film bags
  • Weldable C-Flex and PVC lines
  • Supports high-density T-cell culture (>4 x 106 CD3+ T cells/mL)
  • Supports T-cell activation using Dynabeads magnetic beads, soluble antibodies, and stimulatory antibody-presenting cell protocols
  • Similar phenotype, function (e.g., cytokine secretion profile), and viability to T cells cultured with conventional human AB serum–supplemented medium
  • Supports a T-cell phenotype similar to human serum–supplemented medium
  • Demonstrates enhanced efficacy and persistence of CART-19 cells when grown in medium supplemented with CTS Immune Cell Serum Replacement (ICSR)
Benefits include:
  • Prepare complete medium in a closed system with weldable lines
  • Reduces open steps and contamination risks
  • Easy integration with CTS automated instrumentation
Demonstrated functional performance
Cells grown in CTS OpTmizer T-Cell Expansion SFM with or without human serum demonstrate good cytokine production profiles in polyclonally activated T cells. CTS OpTmizer T-Cell Expansion SFM maintains the desired the CD62L+ central memory T-cell phenotype.
GMP manufactured
The primary manufacturing site for CTS OpTmizer T-Cell Expansion SFM is FDA-registered and has an ISO13485–certified quality management system. The methods and controls used for the manufacturing are in conformity with current Good Manufacturing Practices for medical devices, 21 CFR Part 820, of the regulation.
Versatility
CTS OpTmizer T-Cell Expansion SFM has been widely adopted in clinical T-cell research applications including the culture of CAR T-cells. It works with multiple culture formats: culture flasks, culture bags, and automated and scalable bioreactors. CTS OpTmizer T-Cell Expansion SFM was designed to support expansion of cultured human T-cells without additional serum supplementation. Increased performance is generally observed when supplemented with CTS Immune Cell Serum Replacement.
Specifications
Cell TypeT Cells
Culture EnvironmentBags, Bioreactors
FormatBag
Product LineOpTmizer
Product TypeSerum Free Medium (SFM)
Quantity5 L
Shelf Life12 Months
Shipping ConditionRoom Temperature
ClassificationXeno-free
Culture TypeStatic Culture
FormLiquid
Serum LevelSerum-free
SterilitySterile-filtered
Sterilization Method0.2 μm Sterile-filtered
With AdditivesNone
Without AdditivesNo Phenol Red
Unit SizeEach
Contents & Storage
• 1 x 5000 mL bag CTS OpTmizer Expansion Basal Medium, store at 2°C to 8°C, protect from light
• 1 x 130 mL CTS OpTmizer Expansion Supplement in 250 mL bag, store at 2°C to 8°C, protect from light
Frequently asked questions (FAQs)
Are the 5L and 10L sizes/configurations of CTS OpTmizer SFM, no phenol red (Cat. No. A3705005, A3705007) and CTS OpTmizer Pro SFM (Cat. No. A4966105, A4966107) compatible with CTS devices?
Yes. The 5L and 10L BPCs (bioprocess containers) are weldable to Xenon, Rotea, and DynaCellect consumables.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
With the 5L and 10L sizes of CTS OpTmizer SFM, no phenol red (Cat. No. A3705005, A3705007) and CTS OpTmizer Pro SFM (Cat. No. A4966105, A4966107), how much supplement should be added to the basal medium?
The 5L and 10L kits are designed so that the entire volume of the supplement BPC (bioprocess container) is added to the basal medium BPC.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
With the 5L and 10L sizes of CTS OpTmizer SFM, no phenol red (Cat. No. A3705005, A3705007) and CTS OpTmizer Pro SFM (Cat. No. A4966105, A4966107), how is the supplement added to the basal medium?
The supplement BPC (bioprocess container) has 24 in. of sterile weldable 3/32 in. ID, 5/32 in. OD DHEP-free PVC tubing. With fill volumes of 130 mL and 260 mL for the 5L and 10L kits respectively, the supplement BPC can be directly welded to the basal medium BPC.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What are the differences between the BPCs (bioprocess containers) in the 1L sizes of CTS OpTmizer SFM, no phenol red and CTS OpTmizer Pro SFM vs. the 5L and 10L sizes?
In the 5L and 10L sizes of CTS OpTmizer SFM, no phenol red and CTS OpTmizer Pro SFM, both the basal medium and supplement components are in BPCs (bioprocess containers) composed of Aegis 5-14 film. The basal medium BPC has PVC and C-Flex lines whereas the supplement BPC has a PVC line.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the stability of the CTS OpTmizer SFM, no phenol red and CTS OpTmizer Pro SFM in the new 5L and 10L sizes?
The 5L and 10L sizes of both CTS OpTmizer SFM, no phenol red (Cat. No. A3705005, A3705007) and CTS OpTmizer Pro SFM (Cat. No. A4966105, A4966107) have 12-month expiry.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.