POROS™ CaptureSelect™ KappaXL Affinity HPLC Columns
Thermo Scientific™

POROS™ CaptureSelect™ KappaXL Affinity HPLC Columns

Thermo Scientific™ POROS™ CaptureSelect™ KappaXL Affinity HPLC columns incorporate the advantages of high-speed POROS chromatography resins with the unique selectivity of CaptureSelect affinity ligands.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)
A37063100 mm4.6 mm
A3706130 mm2.1 mm
A3706250 mm4.6 mm
Catalog number A37063
Price (USD)
4,180.00
Each
Length (Metric):
100 mm
Diameter (Metric):
4.6 mm
Price (USD)
4,180.00
Each

POROS CaptureSelect KappaXL Affinity HPLC columns incorporate the advantages of high-speed POROS chromatography resins with the unique selectivity of CaptureSelect™ affinity ligands.

CaptureSelect KappaXL Affinity HPLC columns recognize the Kappa light chain of human immumoglobulin, enabling quantitation and small-scale purification of all immunoglobulins (including IgA, IgM, IgD, IgE), Fab fragments, and bi-specific antibodies containing the Kappa light chain.

Analytical groups supporting biologic drug development require rapid, sensitive, and precise methods for protein quantitation from cell culture, fermentation harvests, or purification intermediates. It is critical for biopharmaceutical manufacturers to be able to monitor bioreactor product titers to determine when to harvest or abort a reactor cycle.

Features of POROS CaptureSelect KappaXL Affinity HPLC columns include:

  • Versitile—applicable for human immunoglobulins, Fab fragments, and bi-specific antibodies containing a Kappa light chain
  • Rapid—POROS resins enable quantitation or purification of immunoglobulins and antibodies in minutes
  • Hands-off operation—HPLC method can be automated for a streamlined operation
  • Robust—designed to deliver high-performance purification over hundreds of chromatography cycles

High speed with high specificity

  • POROS high performance chromatography allows for the quantitation of product from clarified harvest or partially purified intermediates—typically in under three minutes. Moreover, only minimal sample preparation (0.2 μm filtration) is required, and the eluted product is highly purified and suitable for further analysis, including mass spectrometry.

Automation

POROS Affinity HPLC columns are for use with HPLC systems, which allows the automation of chromatography methods, data collection, and data analysis. In addition, the immobilized affinity ligand enables one method and buffer system to be used across a class of molecules, streamlining analytical approaches.

Multiple cycles

Proper sample preparation, adherence to column operating instructions, and the use of an optimized column cleaning regimen results in strong re-use performance. POROS Affinity HPLC columns can be cycled hundreds of times for cost-effective and reproducible quantitation or purification.

Main characteristics

  • Protein binding capacity: 16.5 mg/mL
  • Species specificity: human
  • Column material: PEEK polymer
  • Frit: 2 μm, titanium
  • POROS resin: cross-linked poly(styrene-divinylbenzene)
  • CaptureSelect ligands: single-domain monospecific antibody fragment produced in Saccharomyces cerevisiae
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Binding Capacity16.5 mg/ml
Column FormatAnalytical Column, Biocompatible PEEK
Column TypeAffinity
IncludesColumn includes EZ grip fittings and column test certificate.
Max. Pressure2400 psi (170 bar)
Packing MaterialHighly Crosslinked Styrene-Divinylbenzene polymer (ST-DVB)
Quantity1 Column
Shipping ConditionWet Ice
Stationary PhaseLight Chain Kappa affinity ligand, human‐specific
Temperature2-40°C
Diameter (Metric)4.6 mm
Length (Metric)100 mm
MaterialPEEK
Particle Size20 μm
PhaseAffinity Separation
Pore Size2000 Å
Product LinePOROS CaptureSelect
TechniqueBio-LC
pH1-10
Unit SizeEach