Aliphatic Amine Latex Beads, 2% w/v, 2 μm
Invitrogen™

Aliphatic Amine Latex Beads, 2% w/v, 2 μm

Microspheres (also called latex beads or latex particles) are spherical particles in the colloidal size range that are formed from an amorphous polymer such as polystyrene.
Have Questions?
Catalog NumberQuantity
A3736615 mL
Catalog number A37366
Price (USD)
385.00
Each
Quantity:
15 mL
Price (USD)
385.00
Each

Microspheres (also called latex beads or latex particles) are spherical particles in the colloidal size range that are formed from an amorphous polymer such as polystyrene. Because of the way that the polystyrene chains arrange themselves in the bead, the surface is very hydrophobic in character, making these ideal materials for the adsorption of materials such as proteins.

This aliphatic amine latex has a high density of amine groups attached to the terminus of an aliphatic six-carbon spacer arm. This product can be used to covalently couple proteins. Location of the amine at the end of the spacer arm minimizes difficulties with steric hindrance and conformation since the group has a greater freedom to rotate. In addition, spacer arms improve the kinetics of latex agglutination reactions. The particles are stabilized by the positively-charged amine groups under low to neutral pH conditions. Care should be taken not to use them under high pH.

Microspheres Are Available in Many Sizes and Surface Functionalities

  • 25 sizes to choose from—0.02 microns to 16 microns
  • 20 different surface—for ready conjugation to virtually any material
  • Surfactant free—ultraclean
  • Various quantities—from milliliter to 500-liter amounts

Key Applications of Microspheres

  • Instrument calibration (flow cytometry, microscopy, HTS, HCS)
  • Flow testing (microfluidics, blood flow, water flow, and air flow)
  • Cell biology tracers (cell differentiation and cell tracing)
  • Immunoassays (agglutination tests, ELISA, particle capture, and contrast reagents)

Get More Information

Read a technical overview of microspheres and find out about high activity beads, super active beads, specialty beads, and review bead-coupling protocols on the IDC Surfactant-free Latex Beads application page.

We'll Make a Custom Microsphere Product for You

If you don't see the microsphere you're looking for in our stocked list, we'll make a custom microsphere for you. Our custom conjugation service is efficient and confidential, and we guarantee the quality of our work. We are ISO 13485:2000 certified.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineIDC
Quantity15 mL
Shipping ConditionRoom Temperature
Surface ModificationAliphatic Amine
Diameter (Metric)2 μm
MaterialPolystyrene
Product TypeLatex Beads
Unit SizeEach
Contents & Storage
Store in refrigerator (2°C to 8°C).