MagMAX™ CORE Mechanical Lysis Module & Glass Microbeads Bundle with CORE Chemistry
Applied Biosystems™
MagMAX™ CORE Mechanical Lysis Module & Glass Microbeads Bundle with CORE Chemistry
This MagMAX CORE Mechanical Lysis Module & Glass Microbeads Bundle with CORE Chemistry includes MagMAX CORE Glass Microbeads, a MagMAXRead more
Have Questions?
Catalog NumberQuantity
A37488100 Tests
Catalog number A37488
Price (BRL)
5.073,43
Each
Quantity:
100 Tests
Price (BRL)
5.073,43
Each
This MagMAX CORE Mechanical Lysis Module & Glass Microbeads Bundle with CORE Chemistry includes MagMAX CORE Glass Microbeads, a MagMAX CORE Mechanical Lysis Module, and a MagMAX CORE Nucleic Acid Purification Kit. The Mechanical Lysis Module (Cat. No. A32836) is also available separately.
MagMAX CORE Glass Microbeads are a supplement to the MagMAX CORE Nucleic Acid Purification Kit and MagMAX CORE Mechanical Lysis Module. The beads aid in the homogenization of completely desiccated feces such as environmental samples or fecal pellets.
For Research Use Only
Specifications
For Use With (Application)Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
High-throughput CompatibilityAutomated Protocols, Manual Protocols, Multiplexing
Product LineMagMAX
Quantity100 Tests
Sample TypeBacteria, Blood, Buccal Samples, Cells, DNA, RNA, Tissue, Virus
Shipping ConditionRoom Temperature
TargetTotal RNA, Pathogen DNA/RNA, Genomic DNA, Viral DNA/RNA
Unit SizeEach
Contents & Storage
• 1 x 50 mL MagMAX CORE Glass Microbeads
• 1 x 50 mL MagMAX CORE Lysis Solution
• 1 x 45 mL MagMAX CORE Binding Solution
• 1 x 60 mL MagMAX CORE Wash Solution 1
• 1 x 60 mL MagMAX CORE Wash Solution 2
• 1 x 12 mL MagMAX CORE Elution Buffer
• 1 x 2.2 mL MagMAX CORE Magnetic Beads
• 1 x 1.25 mL MagMAX CORE Proteinase K
• 1 x 45 mL MagMAX CORE Clarifying Solution
• 100 MagMAX CORE Bead Beating Tubes (2 mL)
Frequently asked questions (FAQs)
You offer the MagMAX CORE Mechanical Lysis Module & Glass Microbeads Bundle with CORE Chemistry and the MagMAX CORE Nucleic Acid Purification Kit + Mechanical Lysis Module. What is the difference between these kits and which kit should I order?
The only difference is that the MagMAX CORE Mechanical Lysis Module & Glass Microbeads Bundle with CORE Chemistry (Cat. No. A37488) contains glass microbeads, whereas the other kit (Cat. No. A37487) does not. The beads are also available as a standalone item (Cat. No. A37489) and aid in the homogenization of completely desiccated fecal or environmental samples. These glass microbeads are added to the desiccated sample together with some deionized water, vortexed, and incubated for about 10 minutes prior to the bead beating step.
What is the difference between the bead beating step of the MagMAX Total Nucleic Acid Isolation Kit (Cat. No. AM1840) and the MagMAX CORE Nucleic Acid Purification Kit + Mechanical Lysis Module (Cat. No. A37487)?
The size of the bead tubes that come with these two kits is different, but the bead beating settings are the same. The Mechanical Lysis Module contains a workflow for up to 5 g of feces in addition to the standard 0.3 g protocol.
What type of bacteria should I use the MagMAX CORE Mechanical Lysis Module (Cat. No. A32836) for?
The MagMAX CORE Mechanical Lysis Module is optimized for isolating nucleic acid from difficult to-lyse targets such as mycobacterium (specifically Mycobacterium avium paratuberculosis and tuberculosis), spores, and complex samples used for microbiome testing.