Alexa Fluor™ 790 NHS Ester (Succinimidyl Ester)
Invitrogen™

Alexa Fluor™ 790 NHS Ester (Succinimidyl Ester)

Alexa Fluor™ 790 is a bright and photostable near-IR dye that is spectrally similar to indocyanine green (ICG) and theRead more
Have Questions?
Catalog NumberQuantity
A3756910 mg
Catalog number A37569
Price (EUR)
5.880,00
Each
Quantity:
10 mg
Request bulk or custom format
Price (EUR)
5.880,00
Each
Alexa Fluor™ 790 is a bright and photostable near-IR dye that is spectrally similar to indocyanine green (ICG) and the IRDye™ 800 dye. Used for stable signal generation in imaging and flow cytometry, Alexa Fluor™ 790 dye is water soluble and pH-insensitive from pH 4 to pH 10. Fluorescence of this long-wavelength Alexa Fluor™ dye is not visible to the human eye but is readily detected by most imaging systems. As the longest-wavelength Alexa Fluor™ dye, the emission is well separated from commonly used far-red fluorophores such as Alexa Fluor™ 647 dye or allophycocyanin (APC), facilitating multicolor analysis. This fluorophore is also expected to be useful for small animal in-vivo imaging (SAIVI) applications or for two-color western applications with the LI-COR™ Odyssey™ infrared imaging system. In addition to reactive dye formulations, we offer Alexa Fluor™ 790 dye conjugated to a variety of antibodies, peptides, proteins, tracers, and amplification substrates optimized for cellular labeling and detection.The NHS ester (or succinimidyl ester) of Alexa Fluor™ 790 is the most popular tool for conjugating this dye to a protein or antibody. NHS esters can be used to label to the primary amines (R-NH2) of proteins, amine-modified oligonucleotides, and other amine-containing molecules. The resulting Alexa Fluor™ conjugate will exhibit brighter fluorescence and greater photostability than the conjugates of other spectrally similar fluorophores.

Detailed information about this AlexaFluor™ NHS ester:

Fluorophore label: Alexa Fluor™ 790 dye
Reactive group: NHS ester
Reactivity: Primary amines on proteins and ligands, amine-modified oligonucleotides
Ex/Em of the conjugate: 784/814 nm
Extinction coefficient: 260,000 cm-1M-1
Spectrally similar dyes: indocyanine green (ICG) and the IRDye™ 800 dye
Molecular weight: ∼1750

Typical Conjugation Reaction
You can conjugate amine-reactive reagents with virtually any protein or peptide (the provided protocol is optimized for IgG antibodies). You can scale the reaction for any amount of protein, but the concentration of the protein should be at least 2 mg/mL for optimal results. We recommend trying three different degrees of labeling, using three different molar ratios of the reactive reagent to protein.

The Alexa Fluor™ NHS ester is typically dissolved in high-quality anhydrous dimethylformamide (DMF) or dimethylsulfoxide (DMSO) (D12345), and the reaction is carried out in 0.1–0.2 M sodium bicarbonate buffer, pH 8.3, at room temperature for 1 hour. Because the pKa of the terminal amine is lower than that of the lysine epsilon-amino group, you may achieve more selective labeling of the amine terminus using a buffer closer to neutral pH.

Conjugate Purification
Labeled antibodies are typically separated from free Alexa Fluor™ dye using a gel filtration column, such as Sephadex™ G-25, BioGel™ P-30, or equivalent. For much larger or smaller proteins, select a gel filtration media with an appropriate molecular weight cut-off or purify by dialysis. We offer several purification kits optimized for different quantities of antibody conjugate:
Antibody Conjugate Purification Kit for 0.5-1 mg (A33086)
Antibody Conjugate Purification Kit for 20-50 μg (A33087)
Antibody Conjugate Purification kit for 50-100 μg (A33088)

Learn More About Protein and Antibody Labeling
We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See our Antibody Labeling kits or use our Labeling Chemistry Selection Tool for other choices. To learn more about our labeling kits, read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in The Molecular Probes™ Handbook.

We’ll Make a Custom Conjugate for You
If you can’t find what you’re looking for in our online catalog, we’ll prepare a custom antibody or protein conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Emission814 nm
Excitation784 nm
Product TypeDye
Quantity10 mg
Shipping ConditionRoom Temperature
Product LineAlexa Fluor
Unit SizeEach
Contents & Storage
1 tube
Store dessicated at -20°C, upon reciept