This product is temporarily unavailable. Please contact your Technical Sales Specialist for more information.
Gibco™

L-Leucine, USP/EP/JP

Catalog number:  A37732IN

Back to top

Description

L-Leucine, USP/EP/JP, also known as (2S)-2-Amino-4-methylpentanoic Acid, is a multi-compendial amino acid for your manufacturing needs and part of our Gibco Chemicals family of products.

Gibco Chemicals is a line of raw materials that fulfill needs throughout the drug development process to commercial production. Standard features include cGMP manufacturing, custom specifications, and a variety of package sizes, as well as a portfolio of cGMP supply chain services to help meet your requirements.

Benefits of Gibco Chemicals include:
Access the materials you need for your R&D and manufacturing processes
Realize manufacturing on a global scale with multi-compendial classifications
Maximize supply chain visibility with transparency of manufacturing and distribution
Eliminate need for re-validation with our comprehensive offering of technical documentation
Streamline supply chain operations by outsourcing key needs such as safety stock storage, custom labeling, and poly-palletization

A focus on quality
Gibco Chemicals are manufactured and distributed through ISO-certified, cGMP-compliant facilities to help meet your regulatory needs and mitigate risk.

Transparency of supply chain
Technical documentation, sales support, and technical service teams are available to provide full visibility into product specifications and journey from manufacturer to your hands.

Custom needs are a standard offering
Gibco Chemicals can be customized to meet your unique specifications including custom fill and weigh requirements, testing protocols, and tailgate samples.
For use as a raw material in further manufacturing

Specifications

Chemical Name or Material: L-Leucine, USP/EP/JP
Physical Form: Powder
Product Line: Gibco™ Chemicals
Shipping Condition: Room Temperature

Contents & storage

Contents: amino acid biochemical powder
Storage: ambient temperature in dry, well-ventilated place