AutoLys M TubeLifter
Applied Biosystems™
AutoLys M TubeLifter
The AutoLys M TubeLifter enables a rack of 24 AutoLys M Tubes to be simultaneously “lifted” and readied for centrifugation.자세히 알아보기
Have Questions?
카탈로그 번호수량
A379561 each
카탈로그 번호 A37956
제품 가격(KRW)
4,319,000
온라인 행사
Ends: 31-Dec-2026
5,081,000
할인액 762,000
Each
수량:
1 each
제품 가격(KRW)
4,319,000
온라인 행사
Ends: 31-Dec-2026
5,081,000
할인액 762,000
Each
The AutoLys M TubeLifter enables a rack of 24 AutoLys M Tubes to be simultaneously “lifted” and readied for centrifugation. Once centrifugation is complete the rack of tubes is returned to the TubeLifter and the inner tube of each AutoLys M Tube is separated from the outer tube. The outer tube contains the lysate that is processed further using the MagMAX FFPE DNA/RNA Ultra Kit to recover the nucleic acids.
For Research Use Only. Not for use in diagnostic procedures.
사양
용도(장비)AutoLys TubeLifter System
수량1 each
배송 조건Room Temperature
Unit SizeEach
자주 묻는 질문(FAQ)
Can I use a heat block for the AutoLys M tube incubation?
No. The AutoLys M tubes don't fit in a standard 1.5 mL or 2.0 mL heat block. We recommend incubating the tubes in the tube racks in an incubator or oven.
What kind of centrifuge do I need to spin down the AutoLys M Tubes (Cat. No. A38738)?
Any centrifuge with plate adapters will work. The AutoLys M Tube Racks fit nicely into those adapters.
Do I need to buy the AutoLys M TubeLifter (Cat. No. A37956) or AutoLys M Tube Pliers (Cat. No. A38261) to use the AutoLys M Tubes (Cat. No. A38738)?
No, the AutoLys M Tubes can be lifted manually, but the tools significantly reduce hand strain and increase ease of use as well as throughput.
Do the AutoLys M Tubes come with the MagMAX FFPE DNA/RNA Ultra Kit?
No, they have to be purchased separately together with some other products that are necessary for using the AutoLys M Tube system efficiently:
- AutoLys M Tubes and Caps (Cat. No. A38738)
- Autolys M Tube Locking Lid (Cat. No. A37954)
- AutoLys M TubeLifter (Cat. No. A37956)
- AutoLys M Tube Pliers (Cat. No. A38261)
- AutoLys M Tube Racks (Cat. No. A37955)
How does using the the AutoLys M Tube system help in increase the DNA/RNA yield from FFPE samples?
The FFPE-associated wax and debris are held in the upper chamber of the AutoLys M tube while the lysate passes through. Nucleic acids can then be purified from the clarified lysate. The AutoLys M tubes thus reduce manipulation of the sample eliminating the need to pellet the tissue and wash, so tissue loss is minimized