A549 (+hIGF-1) Control Lysate (1 mg/mL)
Thermo Scientific™

A549 (+hIGF-1) Control Lysate (1 mg/mL)

Thermo Scientific™ A549 (+hIGF-1) Control Lysate is a stimulated cell lysate that has been validated for the expression levels and phosphorylation of key targets in the AKT signaling pathway.
Have Questions?
Catalog NumberQuantity
A379600.5 mL
Catalog number A37960
Price (EUR)
399,33
Online exclusive
459,00
Save 59,67 (13%)
Each
Quantity:
0.5 mL
Price (EUR)
399,33
Online exclusive
459,00
Save 59,67 (13%)
Each
Thermo Scientific™ A549 (+hIGF-1) Control Lysate is a stimulated cell lysate that has been validated for the expression levels and phosphorylation of key targets in the AKT signaling pathway.
  • Validated for the expression of AKT, PTEN, IRS1, IGF1-R, GSK3a, GSK3b, PRAS40, mTOR, p70S6K, and TSC2 proteins by western blotting and mass spectrometry analysis
  • Optimized - stimulation and lysis conditions optimized for detection of AKT signaling targets
  • Convenient - lysate is ready-to-use for immunoprecipitation, immunoprecipitation to mass spectrometry, and western blotting applications
  • Concentration: 1 mg/mL
  • Stable for two years at -80°C
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodMass Spectrometry
Final Product TypeProtein
For Use With (Equipment)Mass Spectrometer
Quantity0.5 mL
Shipping ConditionDry Ice
Starting MaterialCell Lysate
Workflow StepSample Enrichment
FormFrozen Liquid
Product LineThermo Scientific
TypeControl Lysate
Unit SizeEach

Frequently asked questions (FAQs)

What are the characteristics of A549 (+hIGF-1) Control Lysate, and what is its main purpose?

A549 (+hIGF-1) Control Lysate is a stimulated A549 cell lysate that has been validated for the expression levels and phosphorylation of key targets (AKT, PTEN, IRS1, IGF1-R, GSK3a, GSK3b, PRAS40, mTOR, p70S6K, and TSC2) in the AKT signaling pathway. It serves as a positive control for use with SureQuant AKT panel kits.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.