Qualification Services Plan for MiniAmp™ Thermal Cyclers
Actual product may vary
Applied Biosystems™
Qualification Services Plan for MiniAmp™ Thermal Cyclers
Have Questions?
Catalog number A38043
Price (MXN)
-
Qualification services for the MiniAmp Thermal Cyclers are designed to help balance business and laboratory requirements and help you comply with global standards, country-specific regulations, and other laboratory site requirements.
Verify instrument performance and minimize downtime with IQ/OQ services
Our factory-trained and experienced engineers will conduct installation qualification (IQ) and operational qualification (OQ) on the MiniAmp and MiniAmp Plus thermal cyclers in your lab. These qualification services help ensure your instruments are installed, operating, and performing according to manufacturer’s specifications and help move you into production faster. Documentation for audits is provided.
When are IQ/OQ recommended?
• At installation
• After a move
When is OQ recommended?
• After planned maintenance
• After critical repairs
• After additions, changes, or upgrades to the system
• For a scheduled OQ site requirement
For more detailed information, view the figure "Qualification services for MiniAmp Thermal Cyclers" below and visit instrument IQ/OQ/IPV.
Certain exclusions apply. Contact your Services and Support Representative for details at compliancesales@thermofisher.com.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineMiniAmp™
Product SizeOQ, MiniAmp Plus
Unit SizeOQ, MiniAmp Plus