CTS™ AIM-V™ Medium, without phenol red, without antibiotics, 10 L - FAQs
CTS™ AIM-V™ Medium, without phenol red, without antibiotics, 10 L - FAQs
View additional product information for CTS™ AIM-V™ Medium, without phenol red, without antibiotics - FAQs (A3830801, A3830802, A4672701)
7 product FAQs found
What is CTS?
The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.
Will CTS AIM-V Medium without phenol red, without antibiotics (Cat. No. A3830802) be discontinued?
No. CTS AIM-V Medium without phenol red, without antibiotics (Cat. No. A3830802) will not be discontinued. The same catalog number will be available with the new Aegis5-14 film following the planned transition in 2026. 
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What type of film is currently used for packaging CTS AIM-V Medium without phenol red, without antibiotics (Cat. No. A3830802)?
CTS AIM-V Medium without phenol red, without antibiotics (Cat. No. A3830802) is packaged in ASI 28 film, featuring an ethylene vinyl acetate (EVA) contact layer and four spike ports.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the difference between CTS AIM V MINUS VALIDATION Medium (Cat. No. A4000311402) and CTS AIM-V Medium without phenol red, without antibiotics (Cat. No. A3830802)?
The two media have the same formulation and GMP quality and are manufactured in ISO13485, MDSAP, FDA-registered, 21 CFR820 facilities. The main difference lies in the packaging. CTS AIM V MINUS VALIDATION Medium (Cat. No. A4000311402) is packaged in the Aegis 5-14 BioProcessing Container (BPC) with a low-density polyethylene (LDPE) contact layer and updated tubing and connector options. On the other hand, CTS AIM-V Medium without phenol red, without antibiotics (Cat. No. A3830802) is packaged in ASI 28 film with an ethylene vinyl acetate (EVA) contact layer and four spike ports.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Where can I get more information about Aegis 5-14 BPC (BioProcessing Container)?
For additional information regarding Aegis 5-14 BPC, reach out to our Cell Culture Technical Support Team at techsupport@thermofisher.com. 
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the difference between CTS AIM-V Medium and CTS AIM V SFM?
These two media have comparable performance and same cGMP quality. The main difference is that CTS AIM-V Medium (Cat. Nos. A3830801, A3830802, A4672701) does not contain phenol red and antibiotics. This medium formulation has been used as an ancillary reagent in a therapeutic cancer vaccine approved by FDA.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Is CTS AIM-V Medium, without phenol red, without antibiotics ready-to-use?
Yes, CTS AIM-V Medium, without phenol red, without antibiotics is ready-to-use. Additional supplementation with cytokines and growth factors may be required for certain types of cells. Human serum or CTS Immune Cell SR (Cat. Nos. A2596101 and A2596102) can be added to AIM-V medium to promote cell expansion.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.