What does the Ion GeneStudio S5 System with SmartStart include?
The package includes the Ion GeneStudio S5 System and SmartStart orientation. SmartStart orientation is carried out by a Field Application Scientist (FAS) to get you up and running quickly in your lab and reduce downtime. It includes:
- 1 day technology overview and instrument workflow training
- 1 day hands-on application workflow training
- 10 hours bioinformatics consultancy
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
- 1 day technology overview and instrument workflow training
- 1 day hands-on application workflow training
- 10 hours bioinformatics consultancy
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.