Ion Torrent™
Ion GeneStudio™ S5 Plus System Extended Warranty Package
Catalog number: A38410
Ion Torrent™
Ion GeneStudio™ S5 Plus System Extended Warranty Package
Catalog number: A38410
Catalog Number
A38410
Unit Size
System + SmartStart + 3 yr ext warranty
Price (USD)
Catalog Number
Unit Size
System + SmartStart + 1 yr ext warranty
Price (USD)
Catalog NumberUnit SizePrice (USD)
A38410System + SmartStart + 3 yr ext warrantyRequest A Quote
A38409System + SmartStart + 1 yr ext warrantyRequest A Quote
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Ion GeneStudio S5 Plus System is a semiconductor-based next-generation sequencing (NGS) system that enables simple targeted sequencing workflows. From genes to exomes, the Ion GeneStudio S5 Plus System is designed to enable flexibility to help labs manage small and large projects across multiple applications. Combining an extended warranty with your system purchase helps extend the life of your investment.
This package includes the Ion GeneStudio S5 Plus System, SmartStart orientation, and an extended warranty. Key features include:
• Select the chip (Ion 510–550 chip) that fits your application and throughput needs to achieve from 2–130 million reads per run
• Reduce setup time and complexity with cartridge-based plug-and-play reagents
• Complete run and analysis in hours and move forward with your experiments
• Increase efficiency and become operational faster with SmartStart orientation
• Receive fast repair from certified service engineers
• Minimize instrument downtime by troubleshooting with experienced Technical Support teams
For detailed instrument information and specifications, see the Ion GeneStudio S5 Plus System.
Get up and running quickly with SmartStart
SmartStart orientation is carried out by a Field Application Scientist (FAS) to get you up and running quickly in your lab and reduce downtime. SmartStart is the first step of your education journey and is designed to help you work smarter and with greater efficiency. It includes:
• 1-day technology overview and instrument workflow
• 1-day hands-on application workflow training
• 10 hours bioinformatics consultancy
Content will vary depending on your level of knowledge and experience.
Extended warranty plan
Add coverage to your existing manufacturer’s limited warranty with an extended warranty. Each extended warranty includes a planned maintenance visit. Should your instrument require a repair, the extended warranty coverage includes parts, labor, and travel at no additional cost. In addition, you’ll receive:
• On-site guaranteed 2-day response time*
• Scheduled on-site planned maintenance (PM)
• Priority access to Remote Service Engineers
Certain exclusions apply. Contact your Services and Support Representative for details at service.sales@thermofisher.com.
*Availability limited in some geographical areas.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product Line
GeneStudio™
Software Type
Torrent Suite Software, Ion Reporter Software
Shipping Condition
Room Temperature
Package Components
Catalog Number:
A38195
Unit Size:
1 system
Qty:
1
Catalog Number:
TRN00230
Unit Size:
Ion FAS On-Site Training, 1 day
Qty:
1
ZGPCSCBFX10
Catalog Number:
ZGPCSCBFX10
Unit Size:
Qty:
1
A33125
Catalog Number:
A33125
Unit Size:
Qty:
1
ZG61SCIONS5PLUS
Catalog Number:
ZG61SCIONS5PLUS
Unit Size:
Qty:
3