Ion GeneStudio™ S5 Prime System with SmartStart, System + SmartStart - FAQs
Ion GeneStudio™ S5 Prime System with SmartStart, System + SmartStart - FAQs
View additional product information for Ion GeneStudio™ S5 Prime System - FAQs (A38196, A38411)
15 product FAQs found
With Ion S5 2X Cleaning Solution, how long does a "pulse clean" take to complete?
It takes approximately 47 mins to do a "pulse clean",
When using Ion S5 2X Cleaning Solution, where in the software should I select "pulse clean"?
Go to "System tools" and select "pulse clean". The instrument must already have been cleaned after a run.
What are the dimensions of the bottles in Ion S5 2X Cleaning Solution?
Here are the bottle dimensions:
- Length (Metric) 61 mm
- Height (Metric) 131.1 mm
- Width (Metric) 61 mm
How many bottles of Ion S5 2X Cleaning Solution do you provide in Cat. No. A34172?
The kit contains 2 bottles of Ion S5 2X Cleaning Solution.
Which instruments is Ion S5 2X Cleaning Solution compatible with?
Ion S5 2X Cleaning Solution is compatible with GeneStudio S5/GeneStudio S5 Plus/GeneStudio S5 Prime systems and Ion S5/Ion S5 XL systems.
When should I use Ion S5 2X Cleaning Solution?
Ion S5 2X Cleaning Solution is used to perform a pulse clean of your Ion S5 Sequencer (GeneStudio S5/GeneStudio S5 Plus/GeneStudio S5 Prime systems and Ion S5/Ion S5 XL systems) to either recover the instrument performance from a diagnosed solid-state buffer leak or perform preventative maintenance if the instrument has not been used for several months.
What does the Ion GeneStudio S5 Prime System with SmartStart include?
The package includes the Ion GeneStudio S5 Prime System and SmartStart orientation. SmartStart orientation is carried out by a Field Application Scientist (FAS) to get you up and running quickly in your lab and reduce downtime. It includes:
- 1 day technology overview and instrument workflow training
- 1 day hands-on application workflow training
- 10 hours bioinformatics consultancy
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What is the maximum throughput/day for the Ion S5 GeneStudio Prime System?
The Ion S5 GeneStudio Prime System has a maximum throughput/day of 50 Gb (2 Ion 550 Chips).
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How long does it take to run an Ion 550 Chip on the Ion S5 GeneStudio Prime System?
The Ion S5 GeneStudio Prime System takes 8.5 hours to run an Ion 550 Chip.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How long does it take to run an Ion 540 Chip on the Ion S5 GeneStudio Prime System?
The Ion S5 GeneStudio Prime System takes 6.5 hours to run an Ion 540 Chip.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What chips are compatible with the Ion GeneStudio S5 Prime System?
Ion 510 Chip, Ion 520 Chip, Ion 530 Chip, Ion 540 Chip, and Ion 550 Chip are compatible with the Ion GeneStudio S5 Prime System.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Is the Ion GeneStudio S5 Prime System compatible with the Ion One Touch 2 System?
Yes, the Ion GeneStudio S5 Prime System compatible with the Ion One Touch 2 System.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Is the Ion GeneStudio S5 Prime System compatible with the Ion Chef System?
Yes, the Ion GeneStudio S5 Prime System is compatible with the Ion Chef System.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Does the Ion GeneStudio S5 Prime System have a separate server?
Yes, the Ion GeneStudio S5 Prime System has a separate server.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What is the server storage size on the Ion GeneStudio S5 Prime System?
The server storage size is 25 TB.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.