Collibri™ Stranded RNA Library Prep Kit for Illumina™ Systems, with human/mouse/rat rRNA depletion kit and UD indexes, 24 rxns - FAQs
Collibri™ Stranded RNA Library Prep Kit for Illumina™ Systems, with human/mouse/rat rRNA depletion kit and UD indexes, 24 rxns - FAQs
View additional product information for Collibri™ Stranded RNA Library Prep Kit for Illumina™ Systems - FAQs (A39005024, A39003024, A38996096, A39003096, A38994096, A39005096, A38996024, A38994024)
5 product FAQs found
Is the elution buffer, included in the Collibri Stranded RNA Library Prep Kit for Illumina Systems, "Tris buffered" or "Tris-Tween"?
Collibri Stranded RNA Library Prep Kit for Illumina Systems elution buffer is based on Tris-HCl, with no EDTA or EGTA.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Is there an RNA control for optimizing fragment length included with the Collibri Stranded RNA Library Prep Kit for Illumina Systems?
The Collibri Stranded RNA Library Prep Kit for Illumina Systems does not include an RNA control. Recommendations for RNA controls can be found in the Invitrogen Collibri Stranded RNA Library Prep Kit for Illumina User Guide, page 16 "Guidelines for RNA Controls".
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What are the differences between all Collibri Stranded RNA Library Prep Kit for Illumina Systems catalog numbers?
Information regarding the different catalog numbers can be found in the Invitrogen Collibri Stranded RNA Library Prep Kit for Illumina User Guide, page 3.
The different catalog numbers allow for the option to purchase different sizes of the library kit and with or without Unique Dual Indexes.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.