Product Image
Applied Biosystems™
MagMAX™ Saliva gDNA Wash I Solution
This Wash I solution is a component of the MagMAX Saliva gDNA Isolation Kit. It is offered here as aRead more
Have Questions?
Catalog NumberQuantity
A390621.1 L
Catalog number A39062
Price (CLP)
-
Quantity:
1.1 L
This Wash I solution is a component of the MagMAX Saliva gDNA Isolation Kit. It is offered here as a stand-alone reagent for applications that require more Wash I solution than is provided in the kit.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Next-Generation Sequencing, PCR
High-throughput CompatibilityHigh-throughput Compatible, Manual Protocols, Automated Protocols
Product LineMagMAX
Product TypeSaliva gDNA Wash I Solution
Quantity1.1 L
Shipping ConditionRoom Temperature
TargetGenomic DNA
Unit SizeEach
Frequently asked questions (FAQs)
The MagMAX Saliva gDNA Lysis/Binding Solution and Wash I Solution have precipitates in them. Can I still use the buffers?
At lower temperatures, the components of the Lysis/Binding Solution and Wash I Solution may precipitate. To get rid of the precipitate, just warm the solutions to room temperature and invert bottles gently to solubilize the reagents before use.
Can I process less than 200 µL of saliva with the MagMAX Saliva gDNA Isolation Kit?
Yes you can, but the gDNA yields will be very low.
Will the MagMAX Saliva gDNA Isolation Kit work for sample types besides saliva?
No, this kit is specialized for freshly collected saliva and stabilized saliva samples only.
Do you sell the buffers and beads included in the MagMAX Saliva gDNA Isolation Kit as standalone items?
Yes, we offer the following kit components as stand-alone items to accommodate custom workflows or reagent volumes that have been optimized for your specific automation needs:
MagMAX Saliva gDNA Binding Beads: Cat. No. A39061
MagMAX Saliva gDNA Wash I Solution: Cat. No. A39062
MagMAX Saliva gDNA Lysis/Binding Solution: Cat. No. A39063
MagMAX Saliva gDNA Elution Solution: Cat. No. A39064
With the MagMAX Saliva gDNA Isolation Kit, do I use different protocols for processing fresh and stabilized saliva?
No, the same protocols are used for fresh and stabilized saliva.