Invitrogen Collibri Stranded RNA Library Prep Kit

Invitrogen Collibri Stranded RNA Library Prep Kit

The Invitrogen Collibri Stranded RNA Library Preparation Kit Modules are designed for a custom build of next-generation sequencing (NGS) workflow on Illumina systems. The modules originally support whole-transcriptome sequencing (human, mouse, rat) and mRNA sequencing (compatible with all species) applications.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoTipo de productoCantidad
A39117024Kit de preparación de bibliotecas de ARN multifilar24 preparaciones
A39100024Kit de limpieza de biblioteca24 preparaciones
A39114096Kit de limpieza de biblioteca96 preparaciones
A39115024Kit de eliminación de ARNm24 preparaciones
A39115096Kit de eliminación de ARNm96 preparaciones
A39117096Kit de preparación de bibliotecas de ARN multifilar96 preparaciones
A39118024Índices individuales de ARN (set 1-24)24 preparaciones
A39118096Índices individuales de ARN (set 1-96)96 preparaciones
A39119024Índices de ADN UD (set 1-24)24 preparaciones
A39119196Índices de ADN UD (set 1-96)96 preparaciones
Número de catálogo A39117024
Precio (USD)
-
Tipo de producto:
Kit de preparación de bibliotecas de ARN multifilar
Cantidad:
24 preparaciones
The Invitrogen Collibri Stranded RNA Library Prep Kit for Illumina Systems is designed for the robust construction of cDNA libraries for strand-specific RNA sequencing on Illumina next-generation sequencing (NGS) systems. The modules bring together the superior features of SuperScript IV Reverse Transcriptase, Dynabeads magnetic particles, and Platinum SuperFi DNA Polymerase, enabling users to obtain high-quality sequencing-ready libraries.
The Invitrogen Collibri Stranded RNA Library Prep Kit for Illumina Systems is designed for the robust construction of cDNA libraries for strand-specific RNA sequencing on Illumina next-generation sequencing (NGS) systems. The modules bring together the superior features of SuperScript IV Reverse Transcriptase, Dynabeads magnetic particles, and Platinum SuperFi DNA Polymerase, enabling users to obtain high-quality, sequencing-ready libraries.

This Collibri Stranded RNA Library Prep Kit Modules supports fast library preparation within 4.5 hours for mRNA or whole transcriptome libraries with 1–25 ng of mRNA-enriched or ribodepleted RNA sample input. Multiplexing of libraries can be carried out using up to 96 single-indexed or unique dual-indexed primers, enabling single-read or paired-end sequencing. The kit is suitable for various-quality RNA samples, including FFPE samples.

Visual cues of process integrity
For maximum convenience, visual cues of process integrity are included throughout the library preparation workflow. Monitor progress of library generation using visual feedback from dyes in critical reagents. The reaction mix changes color in response to the addition of critical components at each step to ensure that library preparation has the highest chance of success. The inert dyes do not interfere with enzymatic reactions and do not compromise sequencing results.

How it works
Full-length Illumina-compatible adaptors are introduced by PCR amplification using Platinum SuperFi DNA Polymerase to construct single-indexed or unique dual-indexed libraries compatible with single-read or paired-end sequencing. The kit contains 24 or 96 either single-indexed barcoded PCR primers (i7) pre-mixed with a universal i5 primer or unique dual-indexed PCR primers that enable multiplexing of up to 96 libraries. Optimized cleanup steps efficiently remove residual primers and adapter/primer dimers while preserving high library yields.

A related product, the Collibri Library Quantification Kit, is recommended for qPCR-based quantifications of libraries before proceeding to sequencing.

Applications
The Collibri Stranded RNA Library Prep Kit is recommended for:
• Gene expression studies
• Alternative splicing analysis
• Non-coding RNA detection and discovery
• Identification of alternative polyadenylation sites
• Gene fusion detection
• Detection of transcript isoforms

Especificaciones
DescripciónFragmentation, Indexing, Ligation, PCR Amplification, Reverse Transcription
Para utilizar con (aplicación)RNA sequencing; whole transcriptome; mRNA sequencing; small RNA sequencing
Línea de productosCollibri
Tipo de productoKit de preparación de bibliotecas de ARN multifilar
Cantidad24 preparaciones
Tamaño de muestra24 preps
FormatoKit
Unit SizeEach
Contenido y almacenamiento
Almacenar a –20°C