VetMAX™ MastiType Multi Kit
Applied Biosystems™
VetMAX™ MastiType Multi Kit
The VetMAX MastiType Multi Kit enables rapid, accurate detection of 15 mastitis-causing pathogens and one penicillin-resistance gene in bovine milk.詳細を見る
製品番号(カタログ番号)数量
A39227100 tests
製品番号(カタログ番号) A39227
価格(JPY)
344,500
Each
数量:
100 tests
The VetMAX MastiType Multi Kit enables rapid, accurate detection of 15 mastitis-causing pathogens and one penicillin-resistance gene in bovine milk. These 16 markers are detected using real-time PCR amplification of DNA unique to each pathogen in four separate reactions.
The advanced technology of the VetMAX MastiType real-time PCR assays offers these advantages:
• Designed for laboratories requiring high-throughput processing methods
• Faster results—typical time from sample to result is 2.5–3 hours, compared to up to 10 days for a culture
• High accuracy—high sensitivity and specificity for all individual pathogens
• Identifies and quantifies DNA, allowing detection of viable, dead, and growth-inhibited pathogens
• Allows use of preserved milk samples, eliminating the need for cooling during sample transportation
• Can be integrated into milk recording programs using preserved milk
• Validated with convenient MagMAX CORE nucleic acid purification method that does not require aspiration and centrifugation
The VetMAX MastiType Multi assay detects these 15 pathogens and one resistance gene in four separate PCR reactions:
• Staphylococcus aureus
• Staphylococcus spp. (including all major coagulase-negative staphylococci)
• Streptococcus agalactiae
• Streptococcus dysgalactiae
• Streptococcus uberis
• Escherichia coli
• Enterococcus spp. (including E. faecalis and E. faecium)
• Klebsiella oxytoca (and/or K. pneumoniae)
• Serratia marcescens
• Corynebacterium bovis
• Trueperella pyogenes and/or Peptoniphilus indolicus
• Staphylococcal β-lactamase gene (penicillin-resistance gene)
• Mycoplasma bovis
• Mycoplasma spp.
• Yeast
• Prototheca spp.
仕様
使用対象 (装置)7500 Fast System, 7500 System, QuantStudio™
反応数4 Reactions
製品ラインVetMAX
製品タイプPCR Reagents and Detection Kit for Animal Health
数量100 tests
サンプルタイプDNA
出荷条件ドライアイス
検出法Primer-probe
GC-Rich PCR Performance高
PCR法Multiplex qPCR
反応速度高速または標準
Unit SizeEach
組成および保存条件
• 4 x 1.2 mL MastiType Master Mix: contains a Hot Start DNA polymerase in an optimized PCR buffer with magnesium and dNTPs
• 1 x 440 μL MastiType Positive Control: a positive control for the PCR reaction components
• 1 x 550 μL MastiType Multi Primer Mix 1: includes primers for multiplex PCR amplification of Staph. aureus, M. bovis, Enterococcus spp. (incl. E. faecalis and E. faecium), Corynebacterium bovis, and an Internal Amplification Control (IAC)
• 1 x 550 μL MastiType Multi Primer Mix 2: includes primers for multiplex PCR amplification of Staphylococcal ß-lactamase gene (penicillin-resistance gene), E. coli, Strep. dysgalactiae, Mycoplasma spp, and an Internal Amplification Control (IAC)
• 1 x 550 μL MastiType Multi Primer Mix 3: includes primers for multiplex PCR amplification of Staphylococcus spp. (including all major CNS), Streptococcus agalactiae, Streptococcus uberis, Prototheca spp
• 1 x 550 μL MastiType Multi Primer Mix 4: includes primers for multiplex PCR amplification of yeast, Serratia marcescens, Klebsiella oxytoca and/or K. pneumonia, Trueperella pyogenes and/or Peptoniphilus indolicus
Store at -20°C.
よくあるご質問(FAQ)
What kind of milk samples can be tested with the VetMAX MastiType kits?
All VetMAX MastiType kits may be used with milk, bulk milk, frozen milk, or preserved milk (e.g., with bronopol).
What qPCR instrument can I use to run the VetMAX MastiType kits?
Either the Applied Biosystems 7500/7500 Fast Real-Time PCR System or the QuantStudio 5 Real-Time PCR System may be used to run the VetMAX MastiType kits.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Which sample prep kit do you recommend upstream of the VetMAX MastiType kits?
We recommend using the MagMAX CORE Nucleic Acid Purification Kit + MagMAX CORE Mastitis & Panbacteria Module (Cat. No. A40289) with the VetMAX MastiType kits.
Which organisms can be detected by each VetMAX MastiType kit?
The VetMax MastiType Micro4 Kit detects Mycoplasma bovis, Staphylococcus aureus, Streptococcus agalactiae, and Streptococcus uberis.
The VetMax MastiType Myco8 Kit detects Mycoplasma spp., Mycoplasma alkalescens, Mycoplasma bovis, Mycoplasma bovigenitalium, Mycoplasma canadense, Mycoplasma californicum, Staphylococcus aureus, and Streptococcus agalactiae.
The VetMax MastiType Multi Kit detects Staphylococcus aureus, Staphylococcus spp. (including all major coagulase-negative staphylococci), Streptococcus agalactiae, Streptococcus dysgalactiae, Streptococcus uberis, Escherichia coli, Enterococcus spp. (incl. E. faecalis and E. faecium), Klebsiella oxytoca and/or Klebsiella pneumoniae, Serratia marcescens, Corynebacterium bovis, Trueperella pyogenes and/or Peptoniphilus indolicus, Staphylococcal beta-lactamase gene (penicillin-resistance gene), Mycoplasma bovis, Mycoplasma spp., Yeast, and Prototheca spp.