본 제품은 액체 질소로 배송 및 보관되는 세포 관련 제품으로, 해당 제품 구매 시 세포 사용 가이드라인을 반드시 확인해주십시오. 세포사용가이드라인 다운로드
본 제품은 LMO 제품으로, 고객 분께서 LMO 신고 시스템을 통해 직접 수입 신고를 진행해주셔야 합니다. 자세히보기
Expi293F™ GnTI- Cells
Gibco™
Expi293F™ GnTI- Cells
The Expi293F GnTI- cell line is derived from engineered Expi293F cells that do not have N-acetylglucosaminyltransferase I (GnTI) activity and자세히 알아보기
Have Questions?
카탈로그 번호수량
A392401 vial
카탈로그 번호 A39240
제품 가격(KRW)
3,507,000
온라인 행사
Ends: 31-Dec-2026
4,008,000
할인액 501,000
Each
수량:
1 vial
제품 가격(KRW)
3,507,000
온라인 행사
Ends: 31-Dec-2026
4,008,000
할인액 501,000
Each
The Expi293F GnTI- cell line is derived from engineered Expi293F cells that do not have N-acetylglucosaminyltransferase I (GnTI) activity and therefore lack complex N-glycans. It is a powerful tool for the high-yield expression of homogeneously glycosylated recombinant proteins.
Features of Expi293F GnTI- cells include:
• Suspension, high-density culture in Expi293 Expression Medium
• Homogeneous N-glycosylation of expressed proteins
• Equivalent protein yields to parental Expi293F cells—up to 1 g/L
• Significantly higher protein yields than other HEK293 GnTI- cell line alternatives
Frozen cells are supplied in a vial containing 1 mL of cells at 1 x 107 viable cells/mL in Expi293 Expression Medium and 10% DMSO. The cells should be thawed directly into Expi293 Expression Medium.
For Research Use Only. Not for use in diagnostic procedures.
사양
제품라인Expi293
수량1 vial
배송 조건Dry Ice
Unit SizeEach
구성 및 보관
1 vial Expi293F GnTI- Cells (1x107 cells/vial), store in vapor-phase liquid nitrogen
자주 묻는 질문(FAQ)
Can I leave my Expi293F culture expressing for more than 7 days?
The growth and expression characteristics of Expi293F cells are such that, by 7 days post-transfection, the culture medium should be close to being spent and maximal protein expression should have already been achieved. Continued incubation will result in a large decrease in cell culture viability.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
If I let my Expi293F cells grow to a higher density than recommended, will they be okay?
While the Expi293 Expression Medium can support much higher cell densities, we do not recommend growing your Expi293F cultures beyond 5-6 x 10e6 cells/mL, as subsequent transfection and protein expression efficiencies may be reduced. At higher densities, there is also the increased possibility of reaching the point of reduced culture viability. If your seed culture does exceed 5-6 x 10e6 cells/mL, passage them once or twice as detailed in the manual, monitoring them for viability and growth rate. Perform a test transfection using the Protein Expression Control IgG or an expression construct of known yield to determine if cell expression performance has been impacted.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
What method do you recommend for quantifying rabbit IgG expression from pRABBIT IgG IRES-EmGFP Positive Control Vector, Cat. No. A39243?
The formation of intact IgG molecules may be quantified using a sandwich ELISA designed to capture and detect rabbit IgG. Besides the rabbit IgG positive control vector, reagents, and consumables that are included in the kit, you will also need purified rabbit IgG to be used as a standard, F(ab')2-Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, HRP (Cat. No. A10547), Protein A Coated Plates, Clear, 96-Well (Cat. No. 15130), TMB Substrate Kit (Cat. No. 34021), SuperBlock (TBS) Blocking Buffer (Cat. No. 37535), and PBS or TBS buffer for washes. There is an example procedure in our Protein A Coated Plates manual (https://tools.thermofisher.com/content/sfs/manuals/MAN0011310_Thermo Scientific_ProteinA_G_AG_Coat_96Well_UG.pdf). Please note that our R&D scientists determine titer values from crude cell culture supernatants using a Pall Life Sciences FortéBio Octet instrument equipped with a Protein A biosensor.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
Do you offer a GFP-expressing mammalian expression vector that I can use as a control to monitor my transfection and expression with the Expi systems?
We offer pRABBIT IgG IRES-EmGFP Positive Control Vector, Cat. No. A39243, which you can use to monitor your transfection and expression.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
How do I culture Expi293F GnTI-, Expi293F Inducible, or Expi293F Inducible GnTI- cells?
Expi293F GnTI-, Expi293F Inducible, and Expi293F Inducible GnTI- cell lines were derived from the parental Expi293F Cells (Cat. No. A14527) and therefore, share similar morphology and growth characteristics. They are cultured in Expi293 Expression Medium and transfected using ExpiFectamine 293 Transfection Kit following our standard Expi293 protocol.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.