L-Methionine (Methyl-<sup>13</sup>C)
Gibco™

L-Methionine (Methyl-13C)

L-Methionine is a common sulfur-containing amino acid that is commonly used for the isotopic labeling of methionine protein residues forRead more
Have Questions?
Catalog NumberQuantity
A39248225 mg
Catalog number A39248
Price (MXN)
-
Quantity:
225 mg
L-Methionine is a common sulfur-containing amino acid that is commonly used for the isotopic labeling of methionine protein residues for NMR spectroscopy studies. Methionine is a common methyl-group donor to various substrates, such as creatine, epinephrine, ergosterol, and choline.

EC Number: 200-562-9 (unlabeled)
CAS Number: 49705-26-02
Formula: *CH3SCH2CH2CH(NH2)COOH (molecular weight: 150.20)
Synonyms: (S)-2-amino-4-(methylmercapto)butyric acid/L-2-amino-4-(methylthio)cutanoic acid

L-Methionine (Methyl-13C) is supplied in a glass vial containing 225 mg with ≥99% 13C isotopic atom purity.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or Material(S)-2-Amino-4-(Methylmercapto)butyric acid / L-2-Amino-4-(methylthio)cutanoic acid
Physical FormPowder
Recommended StorageStore at -20°C.

Shipping ConditionWet Ice
Molecular Weight (g/mol)150.2
Percent Purity>98%
Quantity225 mg
Unit SizeEach