EZ-Link™ Sulfo-NHS-Biotin, No-Weigh™ Format, 10 x 1 mg - FAQs
EZ-Link™ Sulfo-NHS-Biotin, No-Weigh™ Format, 10 x 1 mg - FAQs
View additional product information for EZ-Link™ Sulfo-NHS-Biotin - FAQs (A39256, 21217)
2 product FAQs found
Can you provide the shelf-life for EZ-Link Sulfo-NHS-Biotin?
EZ-Link Sulfo-NHS-Biotin is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (-20 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
How does a product with "No-Weigh" format reduce my concern over reagent stability?
The No-Weigh product format, where small amounts of reagent are pre-weighed and packaged separately, prevents the contamination and loss of reagent reactivity over time associated with repetitive opening and closing of a single vial. This format enables use of a fresh vial of reagent each time and eliminates the hassle of weighing out small amounts of reagent. Please view additional information and the video at: https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-labeling-crosslinking/no-weigh-packaging.html
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.