Can you provide the shelf-life for Sulfo-NHS (N-hydroxysulfosuccinimide)?
Sulfo-NHS (N-hydroxysulfosuccinimide) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (4 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.