Pierce™ Alkylating Reagents
Pierce™ Alkylating Reagents
Pierce™ Alkylating Reagents
Thermo Scientific™
Pierce™ Alkylating Reagents
Alkylating reagents provided in single-use, pre-weighed formats to streamline protein preparation workflows.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityIncludes
A3927010 x 2 mgChloroacetamide
A3927130 x 9.3 mgIodoacetamide
Catalog number A39270
Price (USD)
103.65
Online exclusive
119.00
Save 15.35
Each
Quantity:
10 x 2 mg
Includes:
Chloroacetamide
Price (USD)
103.65
Online exclusive
119.00
Save 15.35
Each
Thermo Scientific Pierce Iodoacetamide or Chloroacetamide are provided in a no-weigh format, where small amounts of reagent are pre-weighed and packaged for single use. This single-use format prevents the contamination and loss of reagent reactivity over time associated with repetitive opening and closing of a single vial. The format enables use of a fresh vial of reagent each time, and eliminates the hassle of weighing out small amounts of reagent.
Alkylating reagents iodoacetamide and chloracetamide are sulfhydryl-reactive alkylating reagents used to block reduced cysteine residues for protein characterization and peptide mapping. Alkylation with iodoacetamide or chloracetamide after cysteine reduction results in the covalent addition of a carbamidomethyl group (57.07 Da) and prevents the formation of disulfide bonds. Reducing agents added after alkylation will react with excess iodoacetamide.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
IncludesChloroacetamide
Product LinePierce
Quantity10 x 2 mg
Product TypeAlkylating Reagent
Unit SizeEach
Frequently asked questions (FAQs)
How does a product with "No-Weigh" format reduce my concern over reagent stability?
The No-Weigh product format, where small amounts of reagent are pre-weighed and packaged separately, prevents the contamination and loss of reagent reactivity over time associated with repetitive opening and closing of a single vial. This format enables use of a fresh vial of reagent each time and eliminates the hassle of weighing out small amounts of reagent. Please view additional information and the video at: https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-labeling-crosslinking/no-weigh-packaging.html
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.