Ion 510™ & Ion 520™ & Ion 530™ Food Protection Kit – Chef (1 sequencing run per initialization)
Thermo Scientific™

Ion 510™ & Ion 520™ & Ion 530™ Food Protection Kit – Chef (1 sequencing run per initialization)

Perform robust and automated template preparation and sequencing using this kit as part of a complete NGS workflow for Food Authenticity testing.
Have Questions?
Catalog NumberQuantity
A394761 Kit
Catalog number A39476
Price (ZAR)
-
Quantity:
1 Kit

Perform robust and automated template preparation and sequencing using the Ion 510™ & Ion 520™ & Ion 530™ Food Protection Kit – Chef (1 sequencing run per initialization) as a part of Thermo Scientific™ Next Generation Sequencing (NGS) Food Authenticity Workflow.

The Thermo Scientific NGS Food Authenticity Workflow allows identification of the meat, plant and fish species present in even the most challenging sample types; protecting your business from food fraud by verifying the authenticity of samples for improved traceability and food safety.

Identification of the species present in food and feed samples is a critical step for proof of authenticity, verification of origin, traceability of raw materials, and quality control of handling and cleaning processes in food production lines.

The Ion 510™ & Ion 520™ & Ion 530™ Food Protection Kit – Chef (1 sequencing run per initialization) allows identification of meat, fish or plant species present in all food types as a part of a complete NGS Food Authenticity Workflow.

  • Complete: provides template preparation and chip loading reagents
  • Simple: simplifies NGS with ready-to-use, disposable reagent cartridges for sequencing.
  • Rapid: enables fast and simple sequencing instrument set-up with less than 15 minutes hands-on time.
  • Flexible: compatible with the Ion 510, Ion 520, and Ion 530 chips, maximizing flexibility.

The NGS Food Authenticity Workflow utilizes Ion Torrent™ Next Generation Sequencing technology to enable an untargeted screening approach making it possible to identify the species contained in a food sample by comparison with a DNA database of meat, plant or fish species.

The Ion 510 & Ion 520 & Ion 530 Food Protection  Kit-Chef (1 sequencing run per initialization) is used in conjunction with the Ion Chef Food Protection  Instrument and Ion GeneStudio™ S5 Food Protection System to enable automated template preparation and sequencing of up to 400 base-read libraries.

The Ion 510 & Ion 520 & Ion 530 Food Protection Kit contains pre-packaged single-use template and sequencing reagent cartridges for use with the Ion Chef Food Protection System and Ion GeneStudio S5 Food Protection System delivering an automated workflow for sequencing of eight loaded chips with complete run traceability. This kit accommodates one sequencing run per initialization.

For Research Use Only. Not for use in diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionIon 510™ & Ion 520™ & Ion 530™ Food Protection  Kit – Chef (1 sequencing run per initialization)
For Use With (Application)Sequencing
Includes
  • Ion S5 Chef supplies, store at room temperature
  • Ion S5 Chef solutions, store at room temperature
  • Ion 510 & Ion 520 & Ion 530 Chef reagents, store at -5° to -30°C
  • Ion S5 Sequencing solutions, store at room temperature
  • Ion S5 Sequencing reagents, store at -5° to -30°C
Product TypeFood Protection Kit
Quantity1 Kit
FormatKit
Unit SizeEach