Choline Chloride
Gibco™

Choline Chloride

Choline chloride is an essential nutrient commonly found in eggs, chicken, and number of other foods.Applications: choline chloride is aMás información
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
500 g
1 kg
Número de catálogo A39755IP
también denominado A397551P
Precio (MXN)
-
Cantidad:
1 kg
Choline chloride is an essential nutrient commonly found in eggs, chicken, and number of other foods.

Applications: choline chloride is a standard component of cell culture media that serves as a phospholipid precursor for the cells.

Mechanism of action: choline serves as a precursor component to two major cell membrane components: phosphatidylcholine and sphingomyelin. In addition, choline functions as a methyl group donor to several different substrates.

Gibco Chemicals is a line of raw materials that fulfill needs throughout the drug development process to commercial production. Standard features include cGMP manufacturing, custom specifications, and a variety of package sizes, as well as a portfolio of cGMP supply chain services to meet every requirement.

Benefits of Gibco Chemicals include:
• Access the materials you need for your R&D and manufacturing processes
• Realize manufacturing on a global scale with multi-compendial classifications
• Maximize supply chain visibility with transparency of manufacturing and distribution
• Eliminate need for re-validation with comprehensive technical documentation
• Streamline supply chain operations by outsourcing key needs such as safety stock storage, custom labeling, and poly-palletization

A focus on quality
Gibco Chemicals are manufactured and distributed through ISO-certified, cGMP-compliant facilities to meet your regulatory needs and mitigate risk.

Transparency of supply chain
Technical documentation, sales support, and technical service teams are available to provide full visibility into product specifications and the journey from the manufacturer to your hands.

Custom needs are a standard offering
Gibco Chemicals can be customized to meet your unique specifications, including custom fill and weigh requirements, testing protocols, and tailgate samples.

Especificaciones
Almacenamiento recomendadoAlmacenar a temperatura ambiente.
Línea de productosGibco
Cantidad1 kg
Unit SizeEach