TrueMark™ OpenArray™ Urinary Tract Microbiota Comprehensive Plate - FAQs
TrueMark™ OpenArray™ Urinary Tract Microbiota Comprehensive Plate - FAQs
View additional product information for TrueMark™ OpenArray™ Urinary Tract Microbiota Comprehensive Plate - FAQs (A39900)
4 product FAQs found
Can you provide the thermal cycling conditions for the TrueMark OpenArray Urinary Tract Microbiota Comprehensive Plate (Cat. No. A39900)?
No, we cannot provide that information.
However, the experiment was run with a manufacturer-predefined thermal cycling protocol optimized for the panel. Cycling conditions for the OpenArray panels are unique as they pertain to requirements for the technology and are not intended to be replicated for other technologies. Anyone who chooses to replicate these experiments for peer review purposes will have access to the same run protocol even though the details are not visible.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.
Which sample prep kit do you recommend when running urinary tract microbiota profiling experiments?
We recommend the MagMAX DNA Multi-Sample Ultra Kit (Cat. No. A25597) for isolating DNA for urinary tract microbiota profiling experiments.
Do you have any positive controls for the urinary tract microbiota assays?
The TaqMan Urinary Tract Microbiota Amplification Control (Cat. No. A39174) is a linearized multi-target plasmid with target sequences for each available urinary tract microbiota profiling assay.
How can I check for the presence of inhibitors in my samples when running microbiota profiling experiments?
We offer a TaqMan Universal DNA Spike-In Control (Cat. No. A39175 or A39176) that can be added to samples before extraction, to monitor recovery efficiency.