Pierce™ Centrifuge Columns, 34 mL, 25 Columns
Thermo Scientific™

Pierce™ Centrifuge Columns, 34 mL, 25 Columns

Pack these empty 0.5 to 10 mL spin columns with agarose beads to perform small-scale affinity purification methods in 15- or 50-mL conical microcentrifuge tubes.
Catalog NumberQuantity
A4000036425 Columns
Catalog number A40000364
Price (JPY)
19,200
Online offer
Ends: 27-Mar-2026
32,000
Save 12,800 (40%)
25 columns
Quantity:
25 Columns

Pierce Centrifuge Columns

Thermo Scientific Pierce centrifuge columns are convenient tools for handling 40 μL to 20 mL of an affinity support for purification. The columns are made from low-protein-binding polypropylene and contain a polyethylene filter (~30-μm pore size) for compatibility with protein purification.

Simply add the affinity resin to the polypropylene column, remove the twist-off bottom, and allow the resin to pack itself. Then add your sample, allow it to bind to the support, and use a centrifuge to efficiently wash away any contaminants and elute your purified protein.

Pierce centrifuge columns can be used in either spin or gravity-flow format. The spin format reduces sample processing time and makes column washing and elution more efficient. For gravity-flow procedures and binding steps with dilute samples when slow processing time is desired, column extenders can be used to increase the column reservoir capacity. Use any combination of processing techniques to get the best possible chromatography results.

0.8 mL columns

  • Maximum volume: 800 μL
  • Recommended resin volume: 40–400 μL
  • Receiver tube: fits standard microcentrifuge tubes
  • Cap: O-ring screw-top
  • Twist-off bottom

2 mL columns

  • Maximum volume: 5 mL (2 mL resin bed, 3 mL sample)
  • Recommended resin volume: 400 μL–2 mL
  • Receiver tube: fits standard 15 mL conical centrifuge tubes
  • Cap: screw-top
  • Twist-off bottom and press-on cap to reseal

5 mL columns

  • Maximum volume: 8 mL (5 mL resin bed, 3 mL sample)
  • Recommended resin volume: 1–5 mL
  • Receiver tube: fits standard 15 mL conical centrifuge tubes
  • Cap: screw-top
  • Twist-off bottom and press-on cap to reseal

10 mL columns

  • Maximum volume: 22 mL (10 mL resin bed, 12 mL sample)
  • Recommended resin volume: 2–10 mL
  • Receiver tube: fits standard 50 mL conical centrifuge tubes
  • Cap: screw-top
  • Twist-off bottom and press-on cap to reseal

20 mL columns

  • Maximum volume: 34 mL (20 mL resin bed, 14 mL sample)
  • Recommended resin volume: 4–20 mL
  • Receiver tube: fits standard 50 mL conical centrifuge tubes
  • Cap: screw-top
  • Twist-off bottom and press-on cap to reseal

Applications

  • Affinity purification, affinity chromatography
  • Immuno-depletion
  • Ion exchange resins
  • Gel filtration, desalting, buffer exchange
For Research Use Only.
Specifications
Bed Volume20 mL
Column TypeCentrifuge Column
FormatSpin Column
MaterialPolypropylene
Quantity25 Columns
Sample Volume14 mL
TargetBatch Binding
Capacity (Metric)34 mL
Product LinePierce
TypeSpin Column
Unit Size25 columns
Contents & Storage
Room Temperature