Expi293™ PRO Expression Medium
Gibco™

Expi293™ PRO Expression Medium

Expi293 PRO Expression Medium supports rapid, high-yield protein production from HEK293 cells. Designed for scalable, high-density cultures with consistent expression across a variety of proteins, including complex and low-yield targets.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
A400013550420 L
A40001355011 L
A40001355056 x 1 L
A400013550310 L
카탈로그 번호 A4000135504
제품 가격(KRW)
9,009,000
온라인 행사
Ends: 31-Dec-2026
10,597,600
할인액 1,588,600 (15%)
Each
수량:
20 L
제품 가격(KRW)
9,009,000
온라인 행사
Ends: 31-Dec-2026
10,597,600
할인액 1,588,600 (15%)
Each

Gibco Expi293 PRO Expression Medium is a chemically defined, serum-free, protein-free medium for growth and transfection of suspension-adapted human embryonic kidney (HEK) 293 cells. It is specifically designed as a core component to support high density culture of Expi293 PRO cells in the Expi293 PRO Expression System for scalable transient protein expression.

  • Supports growth of suspension HEK 293 cultures to densities of over 15 x 10^6 cells/mL
  • Transfection-compatible serum-free suspension culture medium
  • Helps enable sustained, high level expression of high density transiently transfected cultures, achieving yields of up to 5 g/L of recombinant protein
  • Supports growth and transfection of HEK293 cells in culture formats of 1 mL to 10 L
  • Does not contain phenol red

Expi293 PRO Expression Medium is formulated with GlutaMAX-I reagent. It is ready to use, with no supplementation required. The chemically defined formulation results in high reproducibility and lot-to-lot reliability. Expi293 PRO Expression Medium contains no human or animal-origin components. The medium is not recommended for adherent cell cultures.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
사양
세포주293 (HEK)
세포 유형Embryonic Kidney Cells
제품라인Expi293 PRO
제품 유형Expression Medium
수량20 L
분류Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Culture TypeSuspension Cell Culture
형태Liquid
Serum LevelSerum-free
멸균Sterile
Sterilization MethodSterile-filtered
첨가제 포함GlutaMAX
첨가제 없음No HEPES, No Phenol Red
Unit SizeEach
구성 및 보관
Store in the dark at +2°C to +8°C