OptiPRO™ SFM AGT™ Medium
Gibco™
OptiPRO™ SFM AGT™ Medium
OptiPRO™ SFM AGT™ is a serum free culture medium in a easy-to-use granular format for growth of several kidney derived cell lines MDBK, MDCK, PK-15, and VERO.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
A400016400110 L
A4000164002100 L
Número de catálogo A4000164001
Precio (MXN)
19,956.68
10 L
Cantidad:
10 L
Precio (MXN)
19,956.68
10 L
Gibco OptiPRO SFM AGT Medium is a flexible-format medium that is equivalent to OptiPRO SFM with the addition of Advanced Granulation Technology for simplified preparation. OptiPRO SFM is a serum-free, animal-origin-free culture medium designed for growth of several kidney-derived cell lines, including MDBK, MDCK, PK-15, and VERO, for virus or recombinant protein production. In addition, OptiPRO SFM has been used to successfully grow other cell lines, such as Hela, BHK-21, and COS-7.
Features of OptiPRO SFM AGT Medium include:
  • Serum-free, animal-origin-free, low-protein formulation
  • Easy adaptation for many popular cell lines
  • Scalability in flasks, cell factories, and bioreactors with AGT
Serum-free, animal-origin-free, low-protein formulation
OptiPRO SFM is formulated with a very low protein concentration (< 10 μg/mL), for easier purification of recombinant proteins.
Easy adaptation for many popular cell lines
OptiPRO SFM was designed for growth of kidney-derived cell lines such as MDCK, MDBK, VERO, and PK-15. But it has also been used successfully to grow other cell lines, such as Hela, BHK-21, and COS-7, allowing the use of a single medium for multiple cell lines. OptiPRO SFM is formulated without L-glutamine for greater stability and extended shelf life. Simply add L-glutamine or Glutamax supplement as needed.
cGMP manufacturing and quality system
OptiPRO SFM is manufactured in a cGMP-compliant facility. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
Scalability with Advanced Granulation Technology
Advanced Granulation Technology (AGT) helps simplify medium preparation at large scale with fast rehydration, a just-add-water protocol, and low dust generation, creating less pressure build-up during sterile filtration.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
Línea de célulasBHK-21, Vero, MDCK, MDBK, PK-15, COS-7, HeLa
Tipo de célulaMammalian
Concentración1X
Sistema de expresiónMammalian
FormulaciónProprietary
Tipo de productoMedia
Cantidad10 L
ClasificaciónAnimal Origin, Low Protein, Serum-free, cGMP
Culture TypeAdherent Cell Culture
FormularioPowder
Serum LevelSerum-free
EsterilidadNon-sterile
Sterilization MethodNon-sterile
Con aditivosPhenol Red, Sodium Pyruvate, Sodium Bicarbonate, Thymidine
Sin aditivosNo Glutamine
Unit Size10 L